Skip to Content
Merck
All Photos(1)

Key Documents

EMU034161

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Bst2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAATCTACTTCGCCGTCACAGCGAACAGCGTGGCCTGTAGAGACGGGTTGCGAGCGCAGGCTGAGTGCCGGAACACCACGCACCTGTTGCAGCGCCAGCTCACCCGCACCCAGGACAGTCTGCTGCAGGCCGAGACACAGGCAAACTCCTGCAACCTGACCGTGGTGACCCTTCAGGAGTCCCTGGAGAAGAAGGTGTCTCAAGCCCTGGAGCAGCAGGCCCGCATCAAGGAGCTTGAGAATGAAGTCACGAAGCTGAACCAGGAGCTGGAGAATCTGAGGATCCAAAAGGAGACTTCTAGCACAGTGCAGGTGAACTCTGGCAGCTCCATGGTGGTCTCCAGCCTACTGGTGCTCAAAGTGTCACTGTTCCTGCTCTTTTGAGGACTCATTAGTTGGCAGGTCACAGTTGTTTGAAGTCACTATGGGTCATAGTGACTCTGGAGAGGTCCTGGCAGCCCTGAGGATGTGGAAACCACTAGGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastian Giese et al.
PLoS pathogens, 10(7), e1004189-e1004189 (2014-07-06)
Bst-2/Tetherin inhibits the release of HIV by tethering newly formed virus particles to the plasma membrane of infected cells. Although the mechanisms of Tetherin-mediated restriction are increasingly well understood, the biological relevance of this restriction in the natural target cells
Kerstin Gnirß et al.
Journal of virology, 89(18), 9178-9188 (2015-06-26)
The expression of the antiviral host cell factor tetherin is induced by interferon and can inhibit the release of enveloped viruses from infected cells. The Vpu protein of HIV-1 antagonizes the antiviral activity of tetherin, and tetherin antagonists with Vpu-like
Jaraspim Narkpuk et al.
Biochemical and biophysical research communications, 450(4), 1469-1474 (2014-07-16)
While viral inhibition by tethering of budding virions to host cell membranes has been focused upon as one of the main functions of BST-2/tetherin, BST-2 is thought to possess other functions as well. Overexpression of BST-2 was found here to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service