Skip to Content
Merck
All Photos(1)

Key Documents

EMU020861

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Cd276

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACCAGGATGGAGATGGAGAAGGATCCAAGACAGCTCTACGGCCTCTGAAACCCTCTGAAAACAAAGAAGATGACGGACAAGAAATTGCTTGATTGGGAGCTGCTGCCCTTCCCAGGTGGGGGGCCCACCCTCTGGCAGTGTTGAGCTTCAATGCGAGCCCTTCCCCCCAACGAATGGGTTTGTCCCACAGATCTACCCGTTCGTCAAAGGACGTGGTCCATAGACCACCCACAGCCTTACTTTTCCAATGGACTTAATTCCCATCATCCTGCAGCCTCATTTCTCCAGTGACACGATACACGAACCATCCTGCGGCCTTATTTCCCACGGACACGACACAAAGATGTCCCTCCTCGGTGTTCCTCCAGAGTCGTCTGGTGGCCTTGTGATACGGCGTGAACCTTCTTCCTTCTGCCTTACGTCTAATGGACACACACGCACC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feifei Wang et al.
Cancer investigation, 32(6), 262-271 (2014-05-03)
B7-H3 has been detected in different cancers and correlated to tumor progression and outcome in cancer patients. In this study, we investigated the expression of B7-H3 in tissues and cells of primary hepatocellular carcinoma (PHC) patients. The research showed that
Fu-Biao Kang et al.
Cancer cell international, 15, 45-45 (2015-04-25)
B7-homologue 3 (B7-H3), a recently identified immunoregulatory protein, has been shown to be overexpressed in human hepatocellular carcinoma (HCC). However, whether the dynamic expression pattern of B7-H3 contributes to early invasion of HCC is largely unknown. In addition, the biological
Wei Zhang et al.
OncoTargets and therapy, 8, 1721-1733 (2015-07-24)
The role of B7-H3 in acute monocytic leukemia U937 cells has not been thoroughly investigated. B7-H3 knockdown in the U937 cell line was performed using small hairpin (sh)RNA lentivirus transduction. The effects on cell proliferation, cycle, migration, and invasion were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service