Skip to Content
Merck
All Photos(1)

Key Documents

EMU018761

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Furin

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

AGAGCCAAGAGGGACGTGTATCAGGAGCCCACGGACCCCAAGTTCCCCCAGCAGTGGTACCTGTCTGGTGTCACTCAGCGAGACCTGAATGTGAAGGAGGCCTGGGCCCAGGGCTTCACAGGCCATGGCATTGTGGTCTCCATCCTGGATGACGGCATTGAGAAGAATCATCCCGACCTAGCAGGCAATTATGACCCTGGAGCCAGTTTTGACGTGAATGACCAGGACCCCGACCCACAGCCTCGGTACACACAGATGAATGACAACAGGCATGGCACTCGCTGTGCCGGGGAAGTGGCAGCAGTGGCCAACAATGGTGTCTGTGGCGTAGGTGTAGCTTACAATGCCCGAATTGGAGGGGTGCGGATGTTGGATGGCGAGGTGACTGATGCAGTAGAGGCACGTTCGCTGGGCCTGAATCCCAACCACATCCACAT

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Farhat et al.
British journal of cancer, 122(6), 885-894 (2020-01-29)
Breast cancer is the second most common cancer in the world. Despite advances in therapies, the mechanisms of resistance remain the underlying cause of morbidity and mortality. Lipoic acid (LA) is an antioxidant and essential cofactor in oxidative metabolism. Its
Z Zhou et al.
Cell death & disease, 4, e593-e593 (2013-04-20)
The multinucleated syncytial trophoblast, which forms the outermost layer of the placenta and serves multiple functions, is differentiated from and maintained by cytotrophoblast cell fusion. Deficiencies in syncytial trophoblast differentiation or maintenance likely contribute to intrauterine growth restriction and pre-eclampsia
Xiaokui Yang et al.
PloS one, 8(2), e50479-e50479 (2013-02-19)
Folliculogenesis is tightly controlled by a series of hormones, growth factors and cytokines, many of which are secreted as proproteins and require processing by proteases before becoming functional. Furin is a member of the subtilisin-like proteases that activate large numbers
Jian Shang et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(21), 11727-11734 (2020-05-08)
A novel severe acute respiratory syndrome (SARS)-like coronavirus (SARS-CoV-2) is causing the global coronavirus disease 2019 (COVID-19) pandemic. Understanding how SARS-CoV-2 enters human cells is a high priority for deciphering its mystery and curbing its spread. A virus surface spike
Jian Fu et al.
Molecular carcinogenesis, 54(9), 698-706 (2014-01-18)
Proprotein convertases (PC), a family of serine proteases, process cancer-related substrates such as growth factors, growth factor receptors, cell adhesion molecules, metalloproteinases, etc. HIF-1α is a major transcription factor involved in tumorigenesis by sensing intratumoral hypoxia. Furin (PCSK3) is one

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service