Skip to Content
Merck
All Photos(1)

Key Documents

EMU003031

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Xiap

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCGGGAGCAGCTATCTATCACTTGAGGTCCTGATTGCAGATCTTGTGAGTGCTCAGAAAGATAATACGGAGGATGAGTCAAGTCAAACTTCATTGCAGAAAGACATTAGTACTGAAGAGCAGCTAAGGCGCCTACAAGAGGAGAAGCTTTCCAAAATCTGTATGGATAGAAATATTGCTATCGTTTTTTTTCCTTGTGGACATCTGGCCACTTGTAAACAGTGTGCAGAAGCAGTTGACAAATGTCCCATGTGCTACACCGTCATTACGTTCAACCAAAAAATTTTTATGTCTTAGTGGGGCACCACATGTTATGTTCTTCTTGCTCTAATTGAATGTGTAATGGGAGCGAACTTTAAGTAATCCTGCATTTGCATTCCATTAGCATCCTGCTGTTTCCAAATGG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

mouse ... Xiap(11798)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anak A S S K Dharmapatni et al.
Mediators of inflammation, 2015, 564042-564042 (2015-09-09)
To investigate the effect of Embelin, an inhibitor of X-Linked Inhibitor of Apoptosis Protein (XIAP), on inflammation and bone erosion in a collagen antibody induced arthritis (CAIA) in mice. Four groups of mice (n = 6 per group) were allocated:
Azhar R Hussain et al.
The Journal of clinical endocrinology and metabolism, 100(7), E974-E985 (2015-05-15)
Papillary thyroid cancer (PTC) is the second most common cancer in females in Saudi Arabia. However, the pathogenesis of PTC is still not fully elucidated. To identify potential genes that play important role in progression of PTC, we studied the
N Raulf et al.
British journal of cancer, 111(10), 1955-1964 (2014-10-15)
Current treatment strategies for head and neck cancer are associated with significant morbidity and up to 50% of patients relapse, highlighting the need for more specific and effective therapeutics. Tumour necrosis factor-related apoptosis-inducing ligand (TRAIL) and Smac mimetics (SMs) are
Sojung Park et al.
Anticancer research, 34(7), 3557-3562 (2014-07-02)
Despite the selectivity of Tumor necrosis factor Related Apoptosis-Inducing Ligand (TRAIL) for cancer cell killing activity, breast cancer cells are resistant to TRAIL-induced apoptosis for various reasons. From a functionally-characterized small-molecule dataset, CGP74514A was identified as a TRAIL sensitizer in
Cheng-Yu Tsai et al.
Metallomics : integrated biometal science, 7(11), 1477-1487 (2015-07-25)
Mammalian cells have two influx Cu transporters that form trimers in membranes. CTR1 is the high affinity transporter that resides largely in the plasma membrane, and CTR2 is the low affinity transporter that is primarily associated with vesicular structures inside

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service