Skip to Content
Merck
All Photos(1)

Key Documents

EMU001851

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Nkx3-1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTGGCATCCATCTTTCTGGTAGCAACAGTTCTGCTGATAACTGCATATATCAGAAACTGTCTTCCCTGGGGGTCTCTGGGAAAAAGCCACAGTGGCTGATGTCAAGGAGTCGGAAGCAAGAAATGTACAGATGCAAACTGTTGGGGGTTCCCAGGGAACACTCCAATTCTTCTCTGGTGGGCAGGTGAAAGATCTGGGGCAGTGACTATTTGAAGATGTAGTCCCAGTTCCAAGTGTCCTGTAGGTGACTGCTGCCCGCTCCCCTGTTTAGAGAGAGCCAGGCAAGTTTGAGTCTTGTTTCCAGAACTTAGAATGGCTACAGATGCAATGGCTATATCGTTCCTAGGAATTCTGTTGTGGCAACTCCATCCTCTCCCATGACTGAGTATCATGGAAGGGCCAA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z T Gu et al.
Scientific reports, 5, 11497-11497 (2015-06-25)
In this study, We demonstrated that Bax mitochondrial translocation plays a vital role in the initiation of the mitochondrial signaling pathway upon activation by heat stress. In addition, both p53 mitochondrial translocation and Ca(2+) signal mediated MPTP opening activate Bax
Desheng Zhong et al.
Journal of cellular biochemistry, 115(9), 1624-1635 (2014-05-03)
Pan-Bcl-2 family inhibitor obatoclax has been demonstrated to be effective against various cancers, of which the mechanism of action is not fully understood. In this study, we demonstrate that obatoclax suppressed esophageal cancer cell viability with concomitant G1/G0-phase cell cycle
N Bah et al.
Cell death & disease, 5, e1291-e1291 (2014-06-13)
Antimitotic agents such as microtubule inhibitors (paclitaxel) are widely used in cancer therapy while new agents blocking mitosis onset are currently in development. All these agents impose a prolonged mitotic arrest in cancer cells that relies on sustained activation of
Xi-Liang Wang et al.
PloS one, 10(8), e0136049-e0136049 (2015-08-28)
MicroRNA (miRNA) is a kind of short non-coding RNA, involved in various cellular processes. During keratinocyte differentiation, miRNAs act as important regulators. In this study, we demonstrated by microarray assay that the expression of miR-378b significantly increased during keratinocytes differentiation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service