Skip to Content
Merck
All Photos(1)

Key Documents

EM0100

Sigma-Aldrich

JB-4 Embedding Kit

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€504.00

€504.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€504.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.47

€504.00


Please contact Customer Service for Availability

Request a Bulk Order

application(s)

hematology
histology

Quality Level

storage temp.

room temp

Application

JB-4 is a water-soluble, GMA based, plastic resin kit intended for use in the preparation of embedded samples for high resolution light microscopy. It is a technique now widely used for research and clinical diagnosis. JB-4 yields semi-thin sections (0.5μ-2μ) with excellent morphological preservation.

Kit Components Only

Product No.
Description

  • JB-4 Embedding Solution A (J4954) 800 mL

  • JB-4 Embedding Solution B (J0330) 30 mL

  • Benzoyl Peroxide (J0455) 12 g

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Org. Perox. D - Repr. 1B - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

5.2 - Organic peroxides and self-reacting hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamás Annus et al.
Cell death & disease, 13(4), 363-363 (2022-04-20)
RecQ helicases-also known as the "guardians of the genome"-play crucial roles in genome integrity maintenance through their involvement in various DNA metabolic pathways. Aside from being conserved from bacteria to vertebrates, their importance is also reflected in the fact that
Jessica Sullivan-Brown et al.
Nature protocols, 6(1), 46-55 (2011-01-08)
Histological techniques are critical for observing tissue and cellular morphology. In this paper, we outline our protocol for embedding, serial sectioning, staining and visualizing zebrafish embryos embedded in JB-4 plastic resin-a glycol methacrylate-based medium that results in excellent preservation of
Hui-Min Yin et al.
Science advances, 6(19), eaay9466-eaay9466 (2020-06-05)
The cardiac and hematopoietic progenitors (CPs and HPs, respectively) in the mesoderm ultimately form a well-organized circulation system, but mechanisms that reconcile their development remain elusive. We found that activating transcription factor 3 (atf3) was highly expressed in the CPs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service