Skip to Content
Merck
All Photos(1)

Documents

EHU153841

Sigma-Aldrich

MISSION® esiRNA

targeting human NFKB1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCAGGGTATAGCTTCCCACACTATGGATTTCCTACTTATGGTGGGATTACTTTCCATCCTGGAACTACTAAATCTAATGCTGGGATGAAGCATGGAACCATGGACACTGAATCTAAAAAGGACCCTGAAGGTTGTGACAAAAGTGATGACAAAAACACTGTAAACCTCTTTGGGAAAGTTATTGAAACCACAGAGCAAGATCAGGAGCCCAGCGAGGCCACCGTTGGGAATGGTGAGGTCACTCTAACGTATGCAACAGGAACAAAAGAAGAGAGTGCTGGAGTTCAGGATAACCTCTTTCTAGAGAAGGCTATGCAGCTTGCAAAGAGGCATGCCAATGCCCTTTTCGACTACGCGGTGACAGGAGACGTGAAGATGCTGCTGGCCGTCCAGCGCCATCTCACTGCTGTGCAGGATGAGAATGGGGACAGTGTCTTACACTTAGCAATCATCCACCTTCATTCTCAACTTGTGAGGGATCTACTAGAAGTCACATCTGGTTTGATTTCTGATGACATTATCAACATGAGAAATGATCTGTACCAGACGCCC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Song et al.
Molecular carcinogenesis, 56(1), 36-48 (2016-02-10)
Inflammatory microenvironment created by immune cells is favorable for tumor metastasis. Epithelial-mesenchymal transition (EMT) is involved in the progression of cancer invasion and metastasis in inflammatory microenvironment. In this study, we sought to investigate the effects of Icariside II, a
Zhihong Yuan et al.
Scientific reports, 7, 42028-42028 (2017-02-10)
Triggering receptor expressed on myeloid cells-1(TREM-1) is a member of the superimmunoglobulin receptor family. We have previously shown that TREM-1 prolongs survival of macrophages treated with lipoolysaccharide through Egr2-Bcl2 signaling. Recent studies suggest a role for TREM-1 in viral immunity.
Yunxia Guo et al.
Cytotechnology, 73(1), 115-126 (2021-01-29)
This study intended to investigate the role of NFKB1 in oxidative stress injury and insulin resistance in gestational hypertension (GH) mice. Following establishment of a GH mouse model by high-fat diet, NFKB1, miR-106a, and FLOT2 expression was detected in liver
Jing Zhou et al.
BioMed research international, 2018, 1650456-1650456 (2018-11-08)
Intermittent hypoxia (IH) that resulted from obstructive sleep apnea (OSA) has been found to be a risk factor of coronary artery disease. IH and the receptor for advanced glycation end products (RAGE) expression are known to activate monocyte/macrophage and associated
Shayna E Thomas-Jardin et al.
The Prostate, 80(2), 133-145 (2019-11-16)
The androgen receptor (AR) nuclear transcription factor is a therapeutic target for prostate cancer (PCa). Unfortunately, patients can develop resistance to AR-targeted therapies and progress to lethal disease, underscoring the importance of understanding the molecular mechanisms that underlie treatment resistance.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service