Skip to Content
Merck
All Photos(1)

Documents

EHU136431

Sigma-Aldrich

MISSION® esiRNA

targeting human NRGN

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAAATCCAGGCGAGTTTTCGGGGCCACATGGCGCGGAAGAAGATAAAGAGCGGAGAGCGCGGCCGGAAGGGCCCGGGCCCTGGGGGGCCTGGCGGAGCTGGGGTGGCCCGGGGAGGCGCGGGCGGCGGCCCCAGCGGAGACTAGGCCAGAAGAACTGAGCATTTTCAAAGTTCCCGAGGAGAGATGGATGCCGCGTCCCCTTCGCAGCGACGAGACTTCCCTGCCGTGTTTGTGACCCCCTCCTGCCCAGCAACCTGCCAGCTACAGGAGCCCCCTGCGTCCCAGAGACTCCCTCACCCAGGCAGGCTCCGTCGCGGAGTCGCTGAGTCCGTGCCCTTTTAGTTAGTTCTGCAGTCTAGTATGGTCCCCATTTGCCCTTCCACTCCACCCCACCCTAAACCATGCGCTCCCAATCTTCCTTCTTTTGCTTCTCGC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mariana Marin et al.
Viruses, 11(2) (2019-01-30)
The HIV-1 entry pathway into permissive cells has been a subject of debate. Accumulating evidence, including our previous single virus tracking results, suggests that HIV-1 can enter different cell types via endocytosis and CD4/coreceptor-dependent fusion with endosomes. However, recent studies
Mirko Theis et al.
Journal of biomolecular screening, 20(8), 1018-1026 (2015-04-26)
Broad sequencing enterprises such as the FANTOM or ENCODE projects have substantially extended our knowledge of the human transcriptome. They have revealed that a large portion of genomic DNA is actively transcribed and have identified a plethora of novel transcripts.
Isabel Weinheimer et al.
The Journal of general virology, 95(Pt 2), 486-495 (2013-11-05)
Sweet potato chlorotic stunt virus (SPCSV; genus Crinivirus, family Closteroviridae) causes heavy yield losses in sweet potato plants co-infected with other viruses. The dsRNA-specific class 1 RNase III-like endoribonuclease (RNase3) encoded by SPCSV suppresses post-transcriptional gene silencing and eliminates antiviral
Luca Goitre et al.
Scientific reports, 7(1), 8296-8296 (2017-08-16)
The intracellular scaffold KRIT1/CCM1 is an established regulator of vascular barrier function. Loss of KRIT1 leads to decreased microvessel barrier function and to the development of the vascular disorder Cerebral Cavernous Malformation (CCM). However, how loss of KRIT1 causes the
Pengzhen Wang et al.
Journal of controlled release : official journal of the Controlled Release Society, 279, 220-233 (2018-04-22)
β-site amyloid precursor protein cleaving enzyme 1 (BACE1) is a key enzyme to cleave the amyloid precursor protein to develop Alzheimer's disease (AD). Reducing BACE1 expression in central neuron through RNA interference technology shows great promise to overcome AD. However

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service