Skip to Content
Merck
All Photos(1)

Key Documents

EHU130641

Sigma-Aldrich

MISSION® esiRNA

targeting human SORT1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTCCTGGGTTGGAGATAGCACTGGGGTCATTCTAGTCTTGACTACCTTCCATGTACCACTGGTAATTATGACTTTTGGACAGTCCAAGCTATATCGAAGTGAGGATTATGGGAAGAACTTTAAGGATATTACAGATCTCATCAATAACACCTTTATTCGGACTGAATTTGGCATGGCTATTGGTCCTGAGAACTCTGGAAAGGTGGTGTTAACAGCAGAGGTGTCTGGAGGAAGTCGTGGAGGAAGAATCTTTAGATCATCAGATTTTGCGAAGAATTTTGTGCAAACAGATCTCCCTTTTCATCCTCTCACTCAGATGATGTATAGCCCTCAGAATTCTGATTATCTTTTAGCTCTCAGCACTGAAAATGGCCTGTGGGTGTCCAAGAATTTTGGGGGAAAATGGGAAGAAATCCACAAAGCAGTATGTTTGGCCAAATGGGGATCAGACAACACCATCTTCTTTACAACCTATGCAAATGGCTCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuncheng Lv et al.
Acta biochimica et biophysica Sinica, 51(5), 471-483 (2019-04-06)
Sortilin is closely associated with hyperlipidemia and the risk of atherosclerosis (AS). The role of sortilin and the underlying mechanism in peripheral macrophage are not fully understood. In this study, we investigated the effect of macrophage sortilin on ATP-binding cassette
Swati Venkat et al.
Molecular biology of the cell, 28(19), 2569-2578 (2017-08-05)
Elevated, nontoxic doses of manganese (Mn) protect against Shiga toxin-1-induced cell death via down-regulation of GPP130, a cycling Golgi membrane protein that serves as an endosome-to-Golgi trafficking receptor for the toxin. Mn binds to GPP130 in the Golgi and causes
Keiji Uchiyama et al.
PLoS pathogens, 13(6), e1006470-e1006470 (2017-07-01)
Prion diseases are a group of fatal neurodegenerative disorders caused by prions, which consist mainly of the abnormally folded isoform of prion protein, PrPSc. A pivotal pathogenic event in prion disease is progressive accumulation of prions, or PrPSc, in brains
Hye Youn Sung et al.
Journal of stroke, 20(3), 350-361 (2018-10-13)
The pathogenesis of moyamoya disease (MMD) remains poorly understood, and no reliable molecular biomarkers for MMD have been identified to date. The present study aimed to identify epigenetic biomarkers for use in the diagnosis of MMD. We performed integrated analyses
Fangfang Gao et al.
The American journal of pathology, 190(9), 1931-1942 (2020-06-12)
Pancreatic cancer has a dismal prognosis, and there is no targeted therapy against this malignancy. The neuronal membrane protein sortilin is emerging as a regulator of cancer cell development, but its expression and impact in pancreatic cancer are unknown. This

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service