Skip to Content
Merck
All Photos(1)

Key Documents

EHU091301

Sigma-Aldrich

MISSION® esiRNA

targeting human ZAP70

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGGAAGACGGTGTACCACTACCTCATCAGCCAAGACAAGGCGGGCAAGTACTGCATTCCCGAGGGCACCAAGTTTGACACGCTCTGGCAGCTGGTGGAGTATCTGAAGCTGAAGGCGGACGGGCTCATCTACTGCCTGAAGGAGGCCTGCCCCAACAGCAGTGCCAGCAACGCCTCAGGGGCTGCTGCTCCCACACTCCCAGCCCACCCATCCACGTTGACTCATCCTCAGAGACGAATCGACACCCTCAACTCAGATGGATACACCCCTGAGCCAGCACGCATAACGTCCCCAGACAAACCGCGGCCGATGCCCATGGACACGAGCGTGTATGAGAGCCCCTACAGCGACCCAGAGGAGCTCAAGGACAAGAAGCTCTTCCTGAAGCGCGATAACCTCCTCATAGCTGACATTGAACTTGGCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christina Klasen et al.
Journal of immunology (Baltimore, Md. : 1950), 192(11), 5273-5284 (2014-04-25)
Macrophage migration inhibitory factor (MIF) is a proinflammatory cytokine with chemokine-like functions that plays a pivotal role in the pathogenesis of inflammatory diseases by promoting leukocyte recruitment. We showed that MIF promotes the atherogenic recruitment of monocytes and T cells
Sohye Yoon et al.
PloS one, 10(6), e0126378-e0126378 (2015-06-18)
Adaptive immunity in homeotherms depends greatly on CD4+ Th cells which release cytokines in response to specific antigen stimulation. Whilst bony fish and poikilothermic tetrapods possess cells that express TcR and CD4-related genes (that exist in two forms in teleost

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service