Skip to Content
Merck
All Photos(1)

Key Documents

EHU088121

Sigma-Aldrich

MISSION® esiRNA

targeting human PTBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTTGGCTTCCTTGTGCCTTAAAAAACCTGCCTTCCTGCAGCCACACACCCACCCGGGGTGTCCTGGGGACCCAAGGGGTGGGGGGGTCACACCAGAGAGAGGCAGGGGGCCTGGCCGGCTCCTGCAGGATCATGCAGCTGGGGCGCGGCGGCCGCGGCTGCGACACCCCAACCCCAGCCCTCTAATCAAGTCACGTGATTCTCCCTTCACCCCGCCCCCAGGGCCTTCCCTTCTGCCCCCAGGCGGGCTCCCCGCTGCTCCAGCTGCGGAGCTGGTCGACATAATCTCTGTATTATATACTTTGCAGTTGCAGACGTCTGTGCCTAGCAATATTTCCAGTTGACCAAATATTCTAATCTTTTTTCATTTATATGCAAAAGAAATAGTTTTAAGTAACTTTTTATAGCAAGATGATACAATGGTATGAGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhi-Na Wang et al.
Oncotarget, 8(22), 36185-36202 (2017-04-14)
Polypyrimidine tract-binding protein 1 (PTBP1) involving in almost all steps of mRNA regulation including alternative splicing metabolism during tumorigenesis due to its RNA-binding activity. Initially, we found that high expressed PTBP1 and poor prognosis was interrelated in colorectal cancer (CRC)
Xin Fu et al.
Biochimica et biophysica acta. Molecular cell research, 1865(11 Pt A), 1552-1565 (2018-10-18)
Mesenchymal stem cells (MSCs) hold great promise as attractive vehicles to deliver therapeutic agents against cancer, while the cross-talk between MSCs and cancer cells remains controversial. Here in an indirect co-culture system we observed that MSCs induced the malignancy transformation
Kohei Taniguchi et al.
International journal of molecular sciences, 19(5) (2018-04-27)
Pyruvate kinase is known as the glycolytic enzyme catalyzing the final step in glycolysis. In mammals, two different forms of it exist, i.e., pyruvate kinase M1/2 (PKM) and pyruvate kinase L/R (PKLR). Also, PKM has two isoforms, i.e., PKM1 and
Hoin Kang et al.
The Journal of pathology, 249(3), 395-408 (2019-07-14)
Polypyrimidine tract-binding protein 1 (PTBP1) is one of the most investigated multifunctional RNA-binding proteins (RBP), controlling almost all steps of mRNA metabolism and processing. It has been reported that PTBP1 is overexpressed in many different types of cancer and this
Aline Marnef et al.
Nucleic acids research, 44(3), 1342-1353 (2015-12-15)
Human polypyrimidine tract-binding protein PTB is a multifunctional RNA-binding protein with four RNA recognition motifs (RRM1 to RRM4). PTB is a nucleocytoplasmic shuttle protein that functions as a key regulator of alternative pre-mRNA splicing in the nucleoplasm and promotes internal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service