Skip to Content
Merck
All Photos(1)

Key Documents

EHU080681

Sigma-Aldrich

MISSION® esiRNA

targeting human MAPK9

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAAACCTGTCAGCATTGAAGGAACTCTCACCTCCGTGGGCCTGAAATGCTTGGGAGTTGATGGAACCAAATAGAAAAACTCCATGTTCTGCATGTAAGAAACACAATGCCTTGCCCTACTCAGACCTGATAGGATTGCCTGCTTAGATGATAAAATGAGGCAGAATATGTCTGAAGAAAAAAATTGCAAGCCACACTTCTAGAGATTTTGTTCAAGATCATTTCAGGTGAGCAGTTAGAGTAGGTGAATTTGTTTCAAATTGTACTAGTGACAGTTTCTCATCATCTGTAACTGTTGAGATGTATGTGCATGTGACCACAAATGCTTGCTTGGACTTGCCCATCTAGCACTTTGGAAATCAGTATTTAAATGCCAAATAATCTTCCAGGTAGTGCTGCTTCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leon Caly et al.
Biochemical and biophysical research communications, 483(1), 64-68 (2017-01-08)
Respiratory syncytial virus (RSV) is a major cause of respiratory infections in infants and the elderly, leading to more deaths than influenza each year worldwide. With no RSV antiviral or efficacious vaccine currently available, improved understanding of the host-RSV interaction
Takouhie Mgrditchian et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(44), E9271-E9279 (2017-10-29)
While blocking tumor growth by targeting autophagy is well established, its role on the infiltration of natural killer (NK) cells into tumors remains unknown. Here, we investigate the impact of targeting autophagy gene Beclin1 (BECN1) on the infiltration of NK
Thi Thuy Tien Vo et al.
Journal of Cancer, 11(24), 7253-7263 (2020-11-17)
Recently, ambient air particulate matter (PM) has been shown to increase the risk of oral cancer. The most common malignant tumor in the oral cavity is oral squamous cell carcinoma (OSCC). Recent studies have revealed that surfactin, a cyclic lipopeptide
Kevin Berthenet et al.
Cell reports, 31(10), 107731-107731 (2020-06-11)
Triggering apoptosis remains an efficient strategy to treat cancer. However, apoptosis is no longer a final destination since cancer cells can undergo partial apoptosis without dying. Recent evidence shows that partial mitochondrial permeabilization and non-lethal caspase activation occur under certain
Cuiling Zhong et al.
Nature communications, 11(1), 6330-6330 (2020-12-12)
Endothelial cell (EC) metabolism is thought to be one of the driving forces for angiogenesis. Here we report the identification of the hexosamine D-mannosamine (ManN) as an EC mitogen and survival factor for bovine and human microvascular EC, with an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service