Skip to Content
Merck
All Photos(1)

Key Documents

EHU074291

Sigma-Aldrich

MISSION® esiRNA

targeting human LAMA5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCTACACTGCCCTCAAGTTCTACCTGCAGGGCCCAGAGCCTGAGCCTGGGCAGGGTACCGAGGATCGCTTTGTGATGTACATGGGCAGCCGCCAGGCCACTGGGGACTACATGGGTGTGTCTCTGCGTGACAAGAAGGTGCACTGGGTGTATCAGCTGGGTGAGGCGGGCCCTGCAGTCCTAAGCATCGATGAGGACATTGGGGAGCAGTTCGCAGCTGTCAGCCTGGACAGGACTCTCCAGTTTGGCCACATGTCCGTCACAGTGGAGAGACAGATGATCCAGGAAACCAAGGGTGACACGGTGGCCCCTGGGGCAGAGGGGCTGCTCAACCTGCGGCCAGACGACTTCGTCTTCTACGTCGGGGGGTACCCCAGTACCTTCACGCCCCCTCCCCTGCTTCGCTTCCCCGGCTACCGGGGCTGCATCGAGATGGACACGCTGAATGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana Maltseva et al.
Biochimie, 174, 107-116 (2020-04-26)
The interaction of tumor cells with the extracellular matrix (ECM) may affect the rate of cancer progression and metastasis. One of the major components of ECM are laminins, the heterotrimeric glycoproteins consisting of α-, β-, and γ-chains (αβγ). Laminins interact
Alex Gordon-Weeks et al.
Cancers, 11(5) (2019-05-09)
Hepatic metastatic growth is dependent upon stromal factors including the matrisomal proteins that make up the extracellular matrix (ECM). Laminins are ECM glycoproteins with several functions relevant to tumour progression including angiogenesis. We investigated whether metastatic colon cancer cells produce
Xuemei Zhang et al.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians, 33(7), 1114-1124 (2018-09-12)
Objective: Preeclampsia (PE) is currently thought to associated with oxidative stress and vascular endothelial dysfunction. LAMA5 is associated with the cell migration, proliferation, and vascular endothelial function. The aims of this study are to investigate the expression patterns of LAMA5

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service