Skip to Content
Merck
All Photos(1)

Key Documents

EHU071121

Sigma-Aldrich

MISSION® esiRNA

targeting human NLRP3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ACCTGGAGGATGTGGACTTGAAGAAATTTAAGATGCACTTAGAGGACTATCCTCCCCAGAAGGGCTGCATCCCCCTCCCGAGGGGTCAGACAGAGAAGGCAGACCATGTGGATCTAGCCACGCTAATGATCGACTTCAATGGGGAGGAGAAGGCGTGGGCCATGGCCGTGTGGATCTTCGCTGCGATCAACAGGAGAGACCTTTATGAGAAAGCAAAAAGAGATGAGCCGAAGTGGGGTTCAGATAATGCACGTGTTTCGAATCCCACTGTGATATGCCAGGAAGACAGCATTGAAGAGGAGTGGATGGGTTTACTGGAGTACCTTTCGAGAATCTCTATTTGTAAAATGAAGAAAGATTACCGTAAGAAGTACAGAAAGTACGTGAGAAGCAGATTCCAGTGCATTGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qin Hu et al.
BioFactors (Oxford, England), 44(2), 123-136 (2017-12-02)
Increasing evidence demonstrates that pyroptosis, pro-inflammatory programmed cell death, is linked to atherosclerosis; however, the underlying mechanisms remain to be elucidated. Dihydromyricetin (DHM), a natural flavonoid, was reported to exert anti-oxidative and anti-inflammatory bioactivities. However, the effect of DHM on
Maureen C Ty et al.
EMBO molecular medicine, 11(8), e9903-e9903 (2019-07-03)
Malaria is a highly inflammatory disease caused by Plasmodium infection of host erythrocytes. However, the parasite does not induce inflammatory cytokine responses in macrophages in vitro and the source of inflammation in patients remains unclear. Here, we identify oxidative stress, which
S F Khaiboullina et al.
Scientific reports, 7(1), 16050-16050 (2017-11-24)
ZIKV causes microcephaly by crossing the placental barrier, however, the mechanism of trans-placental dissemination of ZIKV remains unknown. Here, we sought to determine whether monocytes, which can cross tissue barriers, assist ZIKV dissemination to the fetus. We determined this by infecting monocytes
Zhe Lin et al.
Biochimica et biophysica acta. Molecular basis of disease, 1864(9 Pt B), 2890-2900 (2018-06-03)
Oxidative stress and inflammation are closely related to cardiovascular diseases. Although hydrogen sulfide (H2S) has been shown to have powerful anti-oxidative and anti-inflammatory properties, its role in macrophage inflammation was poorly understood. The aim of this study was to investigate
Hui Fu et al.
Frontiers in pharmacology, 9, 968-968 (2018-09-07)
Backgrounds and Aims: Na+ is an important nutrient and its intake, mainly from salt (NaCl), is essential for normal physiological function. However, high salt intake may lead to vascular injury, independent of a rise in blood pressure (BP). Canonical NALP3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service