Skip to Content
Merck
All Photos(1)

Key Documents

EHU070111

Sigma-Aldrich

MISSION® esiRNA

targeting human CCNE1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATCCTCCAAAGTTGCACCAGTTTGCGTATGTGACAGATGGAGCTTGTTCAGGAGATGAAATTCTCACCATGGAATTAATGATTATGAAGGCCCTTAAGTGGCGTTTAAGTCCCCTGACTATTGTGTCCTGGCTGAATGTATACATGCAGGTTGCATATCTAAATGACTTACATGAAGTGCTACTGCCGCAGTATCCCCAGCAAATCTTTATACAGATTGCAGAGCTGTTGGATCTCTGTGTCCTGGATGTTGACTGCCTTGAATTTCCTTATGGTATACTTGCTGCTTCGGCCTTGTATCATTTCTCGTCATCTGAATTGATGCAAAAGGTTTCAGGGTATCAGTGGTGCGACATAGAGAACTGTGTCAAGTGGATGGTTCCATTTGCCATGGTTATAAGGGAGACGGGGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ran Wei et al.
Journal of orthopaedic research : official publication of the Orthopaedic Research Society, 38(9), 1952-1964 (2020-03-13)
While amplified expressed cyclin E1 is a well-known tumorigenic factor and prognostic biomarker in several malignancies, its prognostic predictive potential and function in osteosarcoma is poorly understood. Here we reveal discrete expression pattern, correlation to clinicopathological characteristics and prognosis and
Caishang Zheng et al.
Scientific reports, 7(1), 16422-16422 (2017-11-29)
Enterovirus 71 (EV71) is the predominant causative pathogen of hand-foot-and-mouth disease (HFMD). Contrary to other HFMD-causing enterovirus, EV71 can lead to severe neurological complications, even death. MicroRNAs (miRNAs) are small non-coding RNAs that constitute the largest family of gene regulators
X Zhang et al.
Neoplasma, 66(5), 704-716 (2019-05-28)
Previous studies have reported that miR-107 could be utilized as a potential peripheral biomarker in prostate cancer (PCa). However, the specific functions of miR-107 in prostate cancer and its relevant mechanisms are still unknown. The aim of this research was
Wei-Wei Liu et al.
Cancer management and research, 13, 439-447 (2021-01-28)
To explore the regulatory role of miR497-5p-CCNE1 axis in triple-negative breast cancer (TNBC) cells and its predictive value for early diagnosis. Cancer tissue and adjacent tissue samples were collected from 86 patients with TNBC.RT-PCR was used to detect the expression
Hongqin Wang et al.
Anti-cancer drugs, 30(1), 81-88 (2018-10-03)
Glioma is the most common malignant tumor of the central nervous system with poor survival. Temozolomide (TMZ) is the first-line chemotherapy drug for initial and recurrent glioma treatment with a relatively good efficacy, which exerts its antitumor effects mainly through

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service