Skip to Content
Merck
All Photos(1)

Key Documents

EHU065291

Sigma-Aldrich

MISSION® esiRNA

targeting human NFAT5

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATCCTATTGCCGATGCTCAGAACCTTTCCCAGGAAACTCAAGGTTCTCTCTTTCATAGTCCAAATCCTATTGTCCACAGTCAGACTTCTACAACCTCCTCTGAACAAATGCAGCCTCCAATGTTTCACTCTCAAAGTACCATTGCTGTGTTACAGGGCTCTTCAGTTCCTCAAGACCAGCAGTCAACCAACATATTTCTTTCCCAGAGTCCCATGAATAATCTTCAGACTAACACAGTAGCCCAAGAAGCATTTTTTGCAGCACCGAACTCAATTTCTCCACTTCAGTCAACATCAAACAGTGAACAACAAGCTGCTTTCCAACAGCAAGCTCCAATATCACACATCCAGACTCCTATGCTTTCCCAAGAACAGGCACAACCCCCGCAGCAGGGTTTATTTCAGCCTCAGGTGGCCCTGGGCTCCCTTCCACCTAATCCAATGCCTCAAAGCCAACAAGGAACCAT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saseong Lee et al.
Journal of immunology (Baltimore, Md. : 1950), 201(2), 359-370 (2018-05-26)
Fibroblast-like synoviocytes (FLSs) play a key role in the progression of rheumatoid arthritis (RA) as a primary component of invasive hypertrophied pannus. FLSs of RA patients (RA-FLSs) exhibit cancer-like features, including promigratory and proinvasive activities that largely contribute to joint
Sandrine Herbelet et al.
International journal of molecular sciences, 21(23) (2020-12-09)
Glucocorticoids are drugs of choice in Duchenne muscular dystrophy (DMD), prolonging patients' ambulation. Their mode of action at the protein level is not completely understood. In DMD, muscle tissue is replaced by fibrotic tissue produced by fibroblasts, reducing mobility. Nuclear
Sandrine Herbelet et al.
International journal of molecular sciences, 21(21) (2020-10-30)
Duchenne muscular dystrophy (DMD) is characterized by chronic inflammation and fibrotic tissue production by fibroblasts. The promyogenic factor nuclear factor of activated T-cells 5 (NFAT5) is virtually present in all cells, responding to hyperosmolar or pro-inflammatory stress. In embryogenic fibroblasts
Moritz Veltmann et al.
PloS one, 11(1), e0147312-e0147312 (2016-01-23)
Although systemic hypertension is a risk factor of age-related macular degeneration, antihypertensive medications do not affect the risk of the disease. One condition that induces hypertension is high intake of dietary salt resulting in increased blood osmolarity. In order to
Kunihiko Umekita et al.
Clinical and experimental rheumatology, 37(5), 834-841 (2019-02-16)
Damage-associated molecular patterns (DAMPs) are proposed to drive aberrant stimulation of Toll-like receptors (TLRs) in rheumatoid arthritis (RA) inflamed joints. In the current study we investigated the role of the neutrophil-derived lactoferrin (LTF), as an endogenous ligand for TLR4 in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service