Skip to Content
Merck
All Photos(1)

Key Documents

EHU054231

Sigma-Aldrich

MISSION® esiRNA

targeting human BCL3

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTTCCTTCCTTCCCCATCTCCACCCGCCTTCCTGCCCTTTGCTGGGGTCCTCCGAGGCCCTGGCCGGCCGGTGCCCCCCTCCCCAGCTCCAGGAGGCAGCTGAGGGGGATGGGGGGGCAGATCTTGGACTCATGAGGAGGGGCCCCCCTGCCCTGTGGGGTCAACCCTTCTGGAAACTGTGAAGATCTCACTCTGCCCCCCCCCCCCATCTTCGGGACCAGGATTTGCACAGAAGCACATGCACCTACCCATACACCCCCTCTTCTGAGCACAGATGTTCCCCCATCTCGCTCCCTCCCAGGACTCTGACCCCAGCATTCTCAGGCACCAGTCCCTGTCCGGAATGCCACCCACATCTTCCATTTCCATGTCCCCTCCCAGAGCTGGTGGACCCAGGGAACAGCCACTCCCCTCCACTCTCTACCAGATAACTGAGGAGGGGAGAGGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mikiko Tohyama et al.
European journal of immunology, 48(1), 168-179 (2017-09-14)
IL-22 induces STAT3 phosphorylation and mediates psoriasis-related gene expression. However, the signaling mechanism leading from pSTAT3 to the expression of these genes remains unclear. We focused on Bcl-3, which is induced by STAT3 activation and mediates gene expression. In cultured
SeungPil Yun et al.
Biomolecules & therapeutics, 26(5), 464-473 (2017-08-24)
Cripto is a small glycosylphosphatidylinositol-anchored signaling protein that can detach from the anchored membrane and stimulate proliferation, migration, differentiation, vascularization, and angiogenesis. In the present study, we demonstrated that Cripto positively affected proliferation and survival of mesenchymal stem cells (MSCs)
Xi Chen et al.
Cell death & disease, 7(12), e2508-e2508 (2016-12-03)
Transforming growth factor beta (TGFβ) signaling in breast cancer is selectively associated with pulmonary metastasis. However, the underlying mechanisms remain unclear. Here we show that Bcl-3, a member of the IκB family, serves as a critical regulator in TGFβ signaling
Kangsheng Tu et al.
Oncology reports, 35(4), 2382-2390 (2016-02-18)
Previous studies have demonstrated the aberrant expression and oncogenic role of B-cell CLL/lymphoma-3 (BCL-3) in human malignancies. However, the clinical significance of BCL-3 and its biological function in human hepatocellular carcinoma (HCC) remain unknown. In the present study, the expression
Junhaohui Huo et al.
Acta biochimica et biophysica Sinica, 50(11), 1141-1149 (2018-10-06)
Bcl-3 is an established oncogene in diverse malignant tumors. In this study, we investigated a dual role of Bcl-3 in BL1-subtype triple-negative breast cancer (TNBC). The BL1-subtype TNBC is featured by increasing cell cycle gene expression and the highest sensitivity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service