Skip to Content
Merck
All Photos(1)

Key Documents

EHU053851

Sigma-Aldrich

MISSION® esiRNA

targeting human PROX1

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCTCTCCTTGTCGCTCATAAAGTCCGAGTGCGGCGATCTTCAAGATATGTCTGAAATATCACCTTATTCGGGAAACTCTATGGAGGAAGGATTGTCACCCAATCACTTGAAAAAAGCAAAGCTCATGTTTTTTTATACCCGTTATCCCAGCTCCAATATGCTGAAGACCTACTTCTCCGACGTAAAGTTCAACAGATGCATTACCTCTCAGCTCATCAAGTGGTTTAGCAATTTCCGTGAGTTTTACTACATTCAGATGGAGAAGTACGCACGTCAAGCCATCAACGATGGGGTCACCAGTACTGAAGAGCTGTCTATAACCAGAGACTGTGAGCTGTACAGGGCTCTGAACATGCACTACAATAAAGCAAATGACTTTGAGGTTCCAGAGAGATTCCTGGAAGTTGCTCAGATCACATTACGGGAGTTTTTCAATGCCATTATCGCA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toshihiko Goto et al.
FEBS letters, 591(4), 624-635 (2017-01-28)
Previous reports have revealed that Prospero-related homeobox 1 (Prox1) is required for the migration and differentiation of hepatoblasts during embryonic liver formation. However, the role of Prox1 in adults remains to be elucidated. We created liver-specific Prox1 knockout mice to
Tomonori Sasahira et al.
PloS one, 9(3), e92534-e92534 (2014-03-22)
Prospero homeobox 1 (Prox1) and forkhead box (FOX) C2 regulate angiogenesis and/or lymphangiogenesis. However, the detailed role and function of Prox1 and FOXC2 in cancer remains controversial. In the present study, we examined the expression of Prox1 and FOXC2 proteins
Chang Rae Rho et al.
Investigative ophthalmology & visual science, 56(10), 5871-5879 (2015-09-09)
Prospero homeobox 1 (Prox1) siRNA is a small interfering RNA that is designed to specifically bind Prox1 mRNA. We determined whether Prox1 siRNA inhibits lymphangiogenesis and hemangiogenesis after acute corneal inflammation. Three Prox1 siRNAs were synthesized and investigated for their
Kang-Jin Park et al.
Gastric cancer : official journal of the International Gastric Cancer Association and the Japanese Gastric Cancer Association, 20(1), 104-115 (2016-01-14)
Prospero homeobox 1 (PROX1) functions as a tumor suppressor gene or an oncogene in various cancer types. However, the distinct function of PROX1 in gastric cancer is unclear. We determined whether PROX1 affected the oncogenic behavior of gastric cancer cells
Brian W Wong et al.
Nature, 542(7639), 49-54 (2016-12-27)
Lymphatic vessels are lined by lymphatic endothelial cells (LECs), and are critical for health. However, the role of metabolism in lymphatic development has not yet been elucidated. Here we report that in transgenic mouse models, LEC-specific loss of CPT1A, a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service