Skip to Content
Merck
All Photos(1)

Key Documents

EHU051271

Sigma-Aldrich

MISSION® esiRNA

targeting human PDGFRB

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTGACACTGCACGAGAAGAAAGGGGACGTTGCACTGCCTGTCCCCTATGATCACCAACGTGGCTTTTCTGGTATCTTTGAGGACAGAAGCTACATCTGCAAAACCACCATTGGGGACAGGGAGGTGGATTCTGATGCCTACTATGTCTACAGACTCCAGGTGTCATCCATCAACGTCTCTGTGAACGCAGTGCAGACTGTGGTCCGCCAGGGTGAGAACATCACCCTCATGTGCATTGTGATCGGGAATGAGGTGGTCAACTTCGAGTGGACATACCCCCGCAAAGAAAGTGGGCGGCTGGTGGAGCCGGTGACTGACTTCCTCTTGGATATGCCTTACCACATCCGCTCCATCCTGCACATCCCCAGTGCCGAGTTAGAAGACTCGGGGACCTACACCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary K Goldsmith et al.
Investigative ophthalmology & visual science, 59(11), 4486-4495 (2018-09-08)
Vitreous seeding remains the primary reason for treatment failure in eyes with retinoblastoma (Rb). Systemic and intra-arterial chemotherapy, each with its own inherent set of complications, have improved salvage rates for eyes with advanced disease, but the location and biology
Bhanupriya Madarampalli et al.
Biochimica et biophysica acta. Molecular basis of disease, 1865(6), 1516-1524 (2019-03-17)
Cadherins are homophilic cell-to-cell adhesion molecules that help cells respond to environmental changes. Newly formed cadherin junctions are associated with increased cell phosphorylation, but the pathways driving this signaling response are largely unknown. Since cadherins have no intrinsic signaling activity
N Shioda et al.
Molecular psychiatry, 22(8), 1205-1222 (2016-12-07)
Aberrant dopamine D
Yaoping Liu et al.
Genome research, 25(5), 679-689 (2015-04-11)
Candida albicans, the major invasive fungal pathogen of humans, can cause both debilitating mucosal infections and fatal invasive infections. Understanding the complex nature of the host-pathogen interaction in each of these contexts is essential to developing desperately needed therapies to
Alessia Alunno et al.
Journal of cellular and molecular medicine, 19(7), 1689-1696 (2015-03-11)
It has been recently reported that telocytes, a stromal (interstitial) cell subset involved in the control of local tissue homeostasis, are hampered in the target organs of inflammatory/autoimmune disorders. Since no data concerning telocytes in minor salivary glands (MSGs) are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service