Skip to Content
Merck
All Photos(1)

Key Documents

EHU049601

Sigma-Aldrich

MISSION® esiRNA

targeting human FA2H

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CTCTTCATGCTGGGGACATTCCTCTGGAGCCTCATCGAGTACCTCATCCACCGCTTCCTGTTCCACATGAAGCCCCCCAGCGACAGCTATTACCTCATCATGCTGCACTTCGTCATGCACGGCCAGCACCACAAGGCACCCTTCGACGGCTCCCGCCTGGTCTTCCCCCCTGTGCCAGCCTCCCTGGTGATCGGCGTCTTCTACTTGTGCATGCAGCTCATCCTGCCCGAGGCAGTAGGGGGCACTGTGTTTGCGGGGGGCCTCCTGGGCTACGTCCTCTATGACATGACCCATTACTACCTGCACTTTGGCTCGCCGCACAAGGGCTCCTACCTGTACAGCCTGAAGGCCCACCACGTCAAGCACCACTTTGCACATCAGAAGTCAGGATTTGGTATCAGCACTAAATTGTGGGATTACTGTTTCCACACCCTCACTCCAGAGAAACCCCACCT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masayo Hirao-Suzuki et al.
Biochemical and biophysical research communications, 531(2), 215-222 (2020-08-17)
The functional role of fatty acid 2-hydroxylase (FA2H) is controversial in the field of cancer biology due to the dual role of FA2H, particularly related to its interaction with triple-negative breast cancer (TNBC). A previous biochemical- and clinical-focused study suggested
Bo Hong et al.
Open medicine (Warsaw, Poland), 15(1), 882-889 (2020-12-22)
MicroRNA (miR/miRNA) expression disorders play a crucial role in the development of gastric cancer (GC). Increasing evidence has indicated that miRNAs participate in the process of numerous cancers. Previous research has demonstrated that miR-300 acts as a cancer-promoting factor or
Janani Ravi et al.
Oncotarget, 5(9), 2475-2486 (2014-05-09)
The endocannabinoid anandamide (AEA), a neurotransmitter was shown to have anti-cancer effects. Fatty acid amide hydrolase (FAAH) metabolizes AEA and decreases its anti-tumorigenic activity. In this study, we have analyzed the role of FAAH inhibition in non-small cell lung cancer
Bishuang Gong et al.
Molecular immunology, 122, 173-185 (2020-05-07)
Thymic epithelial cells (TECs) are essential regulators of T cell development and selection. microRNAs (miRNAs) play critical roles in regulating TECs proliferation during thymus involution. miR-205-5p is highly expressed in TECs and increases with age. However, the function and potential

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service