Skip to Content
Merck
All Photos(1)

Key Documents

EHU047831

Sigma-Aldrich

MISSION® esiRNA

targeting human DDR2

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCCCTAATTCTTCCTCCAGCGATGTACGCACTGTCAGTTACACCAATCTGAAGTTTATGGCTACCCAAATTGCCTCTGGCATGAAGTACCTTTCCTCTCTTAATTTTGTTCACCGAGATCTGGCCACACGAAACTGTTTAGTGGGTAAGAACTACACAATCAAGATAGCTGACTTTGGAATGAGCAGGAACCTGTACAGTGGTGACTATTACCGGATCCAGGGCCGGGCAGTGCTCCCTATCCGCTGGATGTCTTGGGAGAGTATCTTGCTGGGCAAGTTCACTACAGCAAGTGATGTGTGGGCCTTTGGGGTTACTTTGTGGGAGACTTTCACCTTTTGTCAAGAACAGCCCTATTCCCAGCTGTCAGATGAACAGGTTATTGAGAATACTGGAGAGTTCTTCCGAGACCAAGGGAGGCAGACTTACCTCCCTCAACCAGCCATTTGTCCTGAC

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Khairul Anam et al.
Stem cell research & therapy, 4(5), 112-112 (2014-01-11)
Knowing the repertoire of cell signaling receptors would provide pivotal insight into the developmental and regenerative capabilities of bone marrow cell (BMC)-derived hematopoietic stem/progenitor cells (HSPCs) and bone marrow mesenchymal stromal cells (BMMSCs). Murine HSPCs were enriched from fluorescence-activated cell
Shijing Jia et al.
American journal of respiratory cell and molecular biology, 59(3), 295-305 (2018-04-14)
Progressive fibrosis is a complication of many chronic diseases, and collectively, organ fibrosis is the leading cause of death in the United States. Fibrosis is characterized by accumulation of activated fibroblasts and excessive deposition of extracellular matrix proteins, especially type
Mereena George Ushakumary et al.
PloS one, 14(12), e0225911-e0225911 (2019-12-06)
Collagen accumulation and remodeling in the vascular wall is a cardinal feature of vascular fibrosis that exacerbates the complications of hypertension, aging, diabetes and atherosclerosis. With no specific therapy available to date, identification of mechanisms underlying vascular fibrogenesis is an
Binhui Xie et al.
Journal of experimental & clinical cancer research : CR, 34, 101-101 (2015-09-13)
Several studies have found that DDR2 is up-regulated in many tumor types and facilitates tumor progression. However, the role of DDR2 in hepatocellular carcinoma (HCC) progression and its downstream signaling pathways remain unclear. DDR2 expression was assessed in several cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service