Skip to Content
Merck
All Photos(1)

Key Documents

EHU047401

Sigma-Aldrich

MISSION® esiRNA

targeting human TFAM

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GATGCTTATAGGGCGGAGTGGCAGGTATATAAAGAAGAGATAAGCAGATTTAAAGAACAGCTAACTCCAAGTCAGATTATGTCTTTGGAAAAAGAAATCATGGACAAACATTTAAAAAGGAAAGCTATGACAAAAAAAAAAGAGTTAACACTGCTTGGAAAACCAAAAAGACCTCGTTCAGCTTATAACGTTTATGTAGCTGAAAGATTCCAAGAAGCTAAGGGTGATTCACCGCAGGAAAAGCTGAAGACTGTAAAGGAAAACTGGAAAAATCTGTCTGACTCTGAAAAGGAATTATATATTCAGCATGCTAAAGAGGACGAAACTCGTTATCATAATGAAATGAAGTCTTGGGAAGAACAAATGATTGAAGTTGGACGAAAGGATCTTCTACGTCGCACAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroya Yamada et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 33(10), 11431-11442 (2019-07-18)
Fructose consumption is rising globally, but maternal high fructose intake might adversely affect offspring. Our previous report demonstrated that excess maternal fructose intake impairs hippocampal function in offspring, indicating that the hippocampi of offspring are highly sensitive to maternal fructose.
Hao Ni et al.
Cancer biology & medicine, 18(1), 139-154 (2021-02-26)
Vascular endothelial growth factor (VEGF), apart from its predominant roles in angiogenesis, can enhance cancer cell proliferation, but its mechanisms remain elusive. The purpose of the present study was therefore to identify how VEGF regulates cancer cell proliferation. VEGF effects
Meng Zhao et al.
Theranostics, 11(4), 1845-1863 (2021-01-08)
Aims: Ischemia-reperfusion injury (IRI)-induced acute kidney injury (IRI-AKI) is characterized by elevated levels of reactive oxygen species (ROS), mitochondrial dysfunction, and inflammation, but the potential link among these features remains unclear. In this study, we aimed to investigate the specific
Xu Jiang et al.
Cancers, 12(2) (2020-02-26)
Mitochondrial transcription factor A (TFAM) is required for mitochondrial DNA replication and transcription, which are essential for mitochondrial biogenesis. Previous studies reported that depleting mitochondrial functions by genetic deletion of TFAM impaired autophagic activities. However, the underlying mechanisms remain largely
A Phillip West et al.
Nature, 520(7548), 553-557 (2015-02-03)
Mitochondrial DNA (mtDNA) is normally present at thousands of copies per cell and is packaged into several hundred higher-order structures termed nucleoids. The abundant mtDNA-binding protein TFAM (transcription factor A, mitochondrial) regulates nucleoid architecture, abundance and segregation. Complete mtDNA depletion

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service