Skip to Content
Merck
All Photos(1)

Documents

EHU043791

Sigma-Aldrich

MISSION® esiRNA

targeting human NUAK2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAGATCGTGATCGTCATGGAGTATGCCAGCCGGGGCGACCTTTATGACTACATCAGCGAGCGGCAGCAGCTCAGTGAGCGCGAAGCTAGGCATTTCTTCCGGCAGATCGTCTCTGCCGTGCACTATTGCCATCAGAACAGAGTTGTCCACCGAGATCTCAAGCTGGAGAACATCCTCTTGGATGCCAATGGGAATATCAAGATTGCTGACTTCGGTCTCTCCAACCTCTACCATCAAGGCAAGTTCCTGCAGACATTCTGTGGGAGCCCCCTCTATGCCTCGCCAGAGATTGTCAATGGGAAGCCCTACACAGGCCCAGAGGTGGACAGCTGGTCCCTGGGTGTTCTCCTCTACATCCTGGTGCATGGCACCATGCCCTTTGATGGGCATGACCATAAGATCCTAGTGAAACAGATCAGCAACGGGGCCTACCGGGAGCCACCTAAACCCTCTGATGCCTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Devon E Mason et al.
The Journal of cell biology, 218(4), 1369-1389 (2019-02-10)
Cell migration initiates by traction generation through reciprocal actomyosin tension and focal adhesion reinforcement, but continued motility requires adaptive cytoskeletal remodeling and adhesion release. Here, we asked whether de novo gene expression contributes to this cytoskeletal feedback. We found that
Takeshi Namiki et al.
Cancer research, 75(13), 2708-2715 (2015-04-03)
The AMPK-related kinase NUAK2 has been implicated in melanoma growth and survival outcomes, but its therapeutic utility has yet to be confirmed. In this study, we show how its genetic amplification in PTEN-deficient melanomas may rationalize the use of CDK2
Mandeep K Gill et al.
Nature communications, 9(1), 3510-3510 (2018-08-31)
In most solid tumors, the Hippo pathway is inactivated through poorly understood mechanisms that result in the activation of the transcriptional regulators, YAP and TAZ. Here, we identify NUAK2 as a YAP/TAZ activator that directly inhibits LATS-mediated phosphorylation of YAP/TAZ
Martin Golkowski et al.
Cell systems, 11(2), 196-207 (2020-08-07)
Hepatocellular carcinoma (HCC) is a complex and deadly disease lacking druggable genetic mutations. The limited efficacy of systemic treatments for advanced HCC implies that predictive biomarkers and drug targets are urgently needed. Most HCC drugs target protein kinases, indicating that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service