Skip to Content
Merck
All Photos(1)

Key Documents

EHU043741

Sigma-Aldrich

MISSION® esiRNA

targeting human AEBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGACACCAGGAGGACTACCCGGTTCACAGGCGTCATCACCCAGGGCAGAGACTCCAGCATCCATGACGATTTTGTGACCACCTTCTTCGTGGGCTTCAGCAATGACAGCCAGACATGGGTGATGTACACCAACGGCTATGAGGAAATGACCTTTCATGGGAACGTGGACAAGGACACACCCGTGCTGAGTGAGCTCCCAGAGCCGGTGGTGGCTCGTTTCATCCGCATCTACCCACTCACCTGGAATGGCAGCCTGTGCATGCGCCTGGAGGTGCTGGGGTGCTCTGTGGCCCCTGTCTACAGCTACTACGCACAGAATGAGGTGGTGGCCACCGATGACCTGGATTTCCGGCACCACAGCTACAAGGACATGCGCCAGCTCATGAAGGTGGTGAACGAGGAGTG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Guo et al.
Behavioural neurology, 2020, 8890452-8890452 (2020-11-24)
Our study was aimed at investigating the mechanistic consequences of the upregulation of adipocyte enhancer-binding protein 1 (AEBP1) in glioblastoma (GBM). The expression of AEBP1 in GBM was assessed by bioinformatics analysis and qRT-PCR; the effects of AEBP1 on GBM
Shouchao Li et al.
Oncology letters, 17(1), 55-62 (2019-01-19)
The present study aimed to analyze adipocyte enhancer-binding protein 1 (AEBP1) expression in colorectal cancer (CRC), with a focus on its possible molecular mechanisms, in order to provide novel insight into the clinical treatment of CRC. Immunohistochemistry (IHC) was used

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service