Skip to Content
Merck
All Photos(1)

Key Documents

EHU030401

Sigma-Aldrich

MISSION® esiRNA

targeting human MELK

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
€192.00
50 μG
€341.00

€192.00


Please contact Customer Service for Availability


Select a Size

Change View
20 μG
€192.00
50 μG
€341.00

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

€192.00


Please contact Customer Service for Availability

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCAAAACCCAAGGGTAACAAGGATTACCATCTACAGACATGCTGTGGGAGTCTGGCTTATGCAGCACCTGAGTTAATACAAGGCAAATCATATCTTGGATCAGAGGCAGATGTTTGGAGCATGGGCATACTGTTATATGTTCTTATGTGTGGATTTCTACCATTTGATGATGATAATGTAATGGCTTTATACAAGAAGATTATGAGAGGAAAATATGATGTTCCCAAGTGGCTCTCTCCCAGTAGCATTCTGCTTCTTCAACAAATGCTGCAGGTGGACCCAAAGAAACGGATTTCTATGAAAAATCTATTGAACCATCCCTGGATCATGCAAGATTACAACTATCCTGTTGAGTGGCAAAGCAAGAATCCTTTTATTCACCTCGATGATGATTGCGTAACAGAACTTTCTGTACATCACAGAAACAACAGGCAAACAATGGAGGATTTAATTTCACTGTGGCAGTATGATCACCTCACGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Cigliano et al.
Medicina (Kaunas, Lithuania), 56(1) (2019-12-22)
Background and Objectives: Intrahepatic cholangiocarcinoma (iCCA) is a pernicious tumor characterized by a dismal outcome and scarce therapeutic options. To substantially improve the prognosis of iCCA patients, a better understanding of the molecular mechanisms responsible for development and progression of
Zhiwei Zhang et al.
Frontiers in oncology, 10, 453-453 (2020-05-12)
Uterine leiomyosarcoma (ULMS) is the most lethal gynecologic malignancy with few therapeutic options. Chemoresistance prevails as a major hurdle in treating this malignancy, yet the mechanism of chemoresistance remains largely unclear. In this study, we certified MELK as a poor
Gang Li et al.
Oncology letters, 15(6), 9934-9940 (2018-05-29)
Maternal embryonic leucine zipper kinase (MELK) is an important regulator in tumorigenesis of human breast cancer, and if silenced leads to programmed cell death in specific breast cancer cell lines, including MDA-MB-231 cells. In the present study, RNA interference, proliferation
Ke Wang et al.
Oncology reports, 39(6), 2777-2786 (2018-04-06)
Retinoblastoma (Rb) is the most frequent primary intraocular tumor usually diagnosed in infants and children, and current therapy for such disease is still limited. Corosolic acid (CA), an ursane-type pentacyclic triterpene, has been assessed as a promising anticancer agent with little impact

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service