Skip to Content
Merck
All Photos(1)

Documents

EHU013081

Sigma-Aldrich

MISSION® esiRNA

targeting human HES1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGTGCTGATAACAGCGGAATCCCCCGTCTACCTCTCTCCTTGGTCCTGGAACAGCGCTACTGATCACCAAGTAGCCACAAAATATAATAAACCCTCAGCACTTGCTCAGTAGTTTTGTGAAAGTCTCAAGTAAAAGAGACACAAACAAAAAATTCTTTTTCGTGAAGAACTCCAAAAATAAAATTCTCTAGAGATAAAAAAAAAAAAAAAAGGAAAATGCCAGCTGATATAATGGAGAAAAATTCCTCGTCCCCGGTGGCTGCTACCCCAGCCAGTGTCAACACGACACCGGATAAACCAAAGACAGCATCTGAGCACAGAAAGTCATCAAAGCCTATTATGGAGAAAAGACGAAGAGCAAGAATAAATGAAAGTCTGAGCCAGCTGAAAACACTGATTTTGGATGCTCTGAAGAAAGATAGCTCGCGG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiadi Wen et al.
Orphanet journal of rare diseases, 8, 100-100 (2013-07-11)
Rare, recurrent genomic imbalances facilitate the association of genotype with abnormalities at the "whole body" level. However, at the cellular level, the functional consequences of recurrent genomic abnormalities and how they can be linked to the phenotype are much less
Xiaoping Du et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 26(5), 1313-1326 (2018-04-24)
Deafness is commonly caused by the irreversible loss of mammalian cochlear hair cells (HCs) due to noise trauma, toxins, or infections. We previously demonstrated that small interfering RNAs (siRNAs) directed against the Notch pathway gene, hairy and enhancer of split 1
Ethan V Abel et al.
PloS one, 9(3), e91983-e91983 (2014-03-22)
Pancreatic cancer stem cells (CSCs) represent a small subpopulation of pancreatic cancer cells that have the capacity to initiate and propagate tumor formation. However, the mechanisms by which pancreatic CSCs are maintained are not well understood or characterized. Expression of
C-R Pradeep et al.
Oncogene, 31(7), 907-917 (2011-07-12)
A large fraction of ductal carcinoma in situ (DCIS), a non-invasive precursor lesion of invasive breast cancer, overexpresses the HER2/neu oncogene. The ducts of DCIS are abnormally filled with cells that evade apoptosis, but the underlying mechanisms remain incompletely understood.
G Fiaschetti et al.
British journal of cancer, 110(3), 636-647 (2013-12-19)
microRNA-9 is a key regulator of neuronal development aberrantly expressed in brain malignancies, including medulloblastoma. The mechanisms by which microRNA-9 contributes to medulloblastoma pathogenesis remain unclear, and factors that regulate this process have not been delineated. Expression and methylation status

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service