Skip to Content
Merck
All Photos(1)

Documents

E8027

Sigma-Aldrich

EphA2/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym(s):

Eck, Myk2, Sek

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

mouse

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

potency

0.078-5 ng/mL

mol wt

monomer calculated mol wt 84 kDa
~105 kDa by SDS-PAGE (reducing)

packaging

pkg of 200 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

ligand binding assay: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Epha2(13836)

General description

EPHA2 (ephrin receptor A2) belongs to the Eph family of receptor tyrosine kinases. It is a type I transmembrane protein, and its exoplasmic domain consists of a ligand binding domain in its N-terminal, a cysteine-rich domain, and two fibronectin type III repeats. Its cytoplasmic domain contains a juxtamembrane region, a tyrosine kinase domain, a SAM (sterile-a-motif) motif, and a PDZ-binding domain.

Biochem/physiol Actions

EPHA2 binds to the ephrin-A ligand and has diverse cellular function. EPHA2 has been shown to be an oncoprotein of importance in a range of cancers. EPHA2 is overexpressed in several human cancer types and promotes malignancy through a mechanism involving RhoA-dependent destabilization of adherens junctions. EPHA2 overexpression induces a FAK-dependent increase in MMP-2 expression and invasiveness and this process that can be reversed by ligation of EPHA2.
Member of the Eph receptor tyrosine kinase family shown to bind ephrin-A1, ephrin-A2, ephrin-A3, ephrin-A4, and ephrin-A5; involved in pattern formation and morphogenesis. EphA2 is expressed on endothelial cells and in tumor xenographs and may regulate tumor neovascularization.

Other Notes

Extracellular domain of mouse EphA2 (amino acids 1-535) fused by means of a polypeptide linker to the Fc region of human IgG1 that is histidine-tagged at the C-terminus.

Physical form

Lyophilized from a 0.2 μm filtered solution in 20 mM Tris, pH 8.0.

Analysis Note

The biological activity is measured by its ability to bind recombinant mouse ephrin-A1/Fc in an ELISA assay.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Duxbury et al.
Biochemical and biophysical research communications, 320(4), 1096-1102 (2004-07-14)
The Eph tyrosine kinases interact with ligands of the Ephrin family and have diverse cellular functions. EphA2 has been recognized to be an oncoprotein of importance in a range of cancers. Here, we examine the effect of EphA2 overexpression and
Wei Bin Fang et al.
Journal of cell science, 121(Pt 3), 358-368 (2008-01-17)
EPHA2 receptor tyrosine kinase is overexpressed in several human cancer types and promotes malignancy. However, the mechanisms by which EPHA2 promotes tumor progression are not completely understood. Here we report that overexpression of a wild-type EPHA2, but not a signaling-defective
Linda M Reis et al.
Molecular vision, 20, 836-842 (2014-06-19)
Congenital cataracts occur in 3-4 per 10,000 live births and account for 5% to 20% of pediatric blindness worldwide. With more than 37 genes known to be associated with isolated congenital cataract, whole exome sequencing (WES) was recently introduced as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service