Skip to Content
Merck
All Photos(1)

Key Documents

E7904

Sigma-Aldrich

Erythromycin

Biotechnology Performance Certified

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H67NO13
CAS Number:
Molecular Weight:
733.93
Beilstein:
75279
EC Number:
MDL number:
UNSPSC Code:
51101500
PubChem Substance ID:

grade

Biotechnology Performance Certified

form

solid

impurities

endotoxin, tested

color

white

mp

133 °C

solubility

ethanol: soluble

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

Mode of action

protein synthesis | interferes

SMILES string

CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O

InChI

1S/C37H67NO13/c1-14-25-37(10,45)30(41)20(4)27(39)18(2)16-35(8,44)32(51-34-28(40)24(38(11)12)15-19(3)47-34)21(5)29(22(6)33(43)49-25)50-26-17-36(9,46-13)31(42)23(7)48-26/h18-26,28-32,34,40-42,44-45H,14-17H2,1-13H3/t18-,19-,20+,21+,22-,23+,24+,25-,26+,28-,29+,30-,31+,32-,34+,35-,36-,37-/m1/s1

InChI key

ULGZDMOVFRHVEP-RWJQBGPGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: macrolide

Application

Erythromycin is an antibiotic produced by growth of certain strains of Streptomyces erythreus. This product is composed largely of erythromycin A with small amounts of erythromycins B and C and is recommended for concentration at 100 mg/L. Concentrations between 50 and 200 mg/L have also proven effective in controlling bacterial growth. Erythromycin has been used as a motilin receptor agonist, to block respiratory glycoconjugate secretion in human airways in vitro, and for selecting plasmid-cured and recombinant lactococcus lactis MG1363 strains.

Biochem/physiol Actions

Mode of Action: Erythromycin acts by inhibiting elongation at the transpeptidation step, specifically aminoacyl translocation from the A-site to P-site by binding to the 50s subunit of the bacterial 70s rRNA complex.

Antimicrobial Spectrum: This product acts against both gram-negative and gram-positive bacteria.

Caution

This product is stable in solution at 37°C for 3 days. Stock solutions should be stored at 2-8°C.

Preparation Note

This product is soluble in water at 2 mg/mL, with a 0.067% solution in water yielding a pH of 8.0-10.5. It is also soluble in ethanol at 50 mg/mL, yielding a clear, colorless to faint yellow solution. It is freely soluble in alcohol, acetone, chloroform, acetonitrile and ethyl acetate but forms salts with acids. All solutions should be protected from light.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Czarnetzki et al.
JAMA surgery, 150(8), 730-737 (2015-06-18)
Patients undergoing emergency procedures under general anesthesia have impaired gastric emptying and are at high risk for aspiration of gastric contents. Erythromycin has strong gastric prokinetic properties. To evaluate the efficacy of erythromycin lactobionate in gastric emptying in patients undergoing
Ying Ying Chan et al.
Antimicrobial agents and chemotherapy, 51(2), 623-630 (2006-12-06)
The gram-negative soil bacillus Burkholderia pseudomallei is the causative agent of melioidosis, a severe and potentially fatal septicemic disease that is endemic to Southeast Asia and northern Australia. Its intrinsic resistance to many antibiotics is attributed mainly to the presence
Leena Keurulainen et al.
Journal of medicinal chemistry, 53(21), 7664-7674 (2010-10-12)
Chlamydia pneumoniae is an intracellular bacterium that responds poorly to antibiotic treatment. Insufficient antibiotic usage leads to chronic infection, which is linked to disease processes of asthma, atherosclerosis, and Alzheimer's disease. The Chlamydia research lacks genetic tools exploited by other
David J Serisier et al.
JAMA, 309(12), 1260-1267 (2013-03-28)
Macrolide antibiotics such as erythromycin may improve clinical outcomes in non-cystic fibrosis (CF) bronchiectasis, although associated risks of macrolide resistance are poorly defined. To evaluate the clinical efficacy and antimicrobial resistance cost of low-dose erythromycin given for 12 months to
Vanessa G Allen et al.
Antimicrobial agents and chemotherapy, 55(2), 703-712 (2010-11-26)
Surveillance of gonococcal antimicrobial resistance and the molecular characterization of the mechanisms underlying these resistance phenotypes are essential in order to establish correct empirical therapies, as well as to describe the emergence of new mechanisms in local bacterial populations. To

Articles

Antibiotics targeting bacterial ribosomes disrupt protein synthesis, a key process in bacterial growth inhibition.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service