Skip to Content
Merck
All Photos(1)

Key Documents

E6500

Sigma-Aldrich

Ergonovine maleate salt

Synonym(s):

Ergometrine maleate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H23N3O2 · C4H4O4
CAS Number:
Molecular Weight:
441.48
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

storage temp.

−20°C

SMILES string

OC(=O)\C=C/C(O)=O.C[C@@H](CO)NC(=O)[C@H]1CN(C)[C@@H]2Cc3c[nH]c4cccc(C2=C1)c34

InChI

1S/C19H23N3O2.C4H4O4/c1-11(10-23)21-19(24)13-6-15-14-4-3-5-16-18(14)12(8-20-16)7-17(15)22(2)9-13;5-3(6)1-2-4(7)8/h3-6,8,11,13,17,20,23H,7,9-10H2,1-2H3,(H,21,24);1-2H,(H,5,6)(H,7,8)/b;2-1-/t11-,13+,17+;/m0./s1

InChI key

YREISLCRUMOYAY-IIPCNOPRSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Oxytocic, potent antagonist at smooth muscle serotonergic receptors.
Oxytocic, stimulates contraction of uterine muscle; partial agonist at serotonergic and dopaminergic receptors in CNS; selective, potent antagonist at smooth muscle serotonergic receptors.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tsuyoshi Ito et al.
Circulation. Cardiovascular imaging, 5(2), 226-232 (2012-02-22)
The potential of multislice CT (MSCT) to predict coronary spasm has not been elucidated. The aim of this study was to investigate whether the morphological features observed on MSCT at the site of ergonovine-induced epicardial spasm could be used as
Wayne Siegel
The American journal of cardiology, 105(8), 1198-1199 (2010-04-15)
This article provides narration of the events during the development of ergonovine maleate infusions for detecting coronary artery spasm at the Cleveland Clinic in 1973. This was the first safe, reproduceable and reliable test for angiographic visualization of the pathophysiology
Rany Harara et al.
The journal of obstetrics and gynaecology research, 37(9), 1203-1207 (2011-04-27)
The aim of this work was to compare the effect of intraumbilical injection of three different uterotonic solutions in the management of retained placenta. This study was conducted in Ain-Shams University Maternity Hospital, Cairo, Egypt. A total of 78 women
Active management of the third stage of labour.
Nasreen Aflaifel et al.
BMJ (Clinical research ed.), 345, e4546-e4546 (2012-07-10)
Hosam E Matar et al.
BMC pregnancy and childbirth, 10, 32-32 (2010-06-24)
Care for women during the third stage aims to reduce the risk of major haemorrhage, but is very variable. The current World Health Organisation (WHO) recommendation is that care should include administration of a uterotonic (oxytocin, if it is available)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service