All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H8N4S
CAS Number:
Molecular Weight:
180.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
Recommended Products
form
solid
SMILES string
[H]n1cnc2c(SCC)ncnc12
InChI
1S/C7H8N4S/c1-2-12-7-5-6(9-3-8-5)10-4-11-7/h3-4H,2H2,1H3,(H,8,9,10,11)
InChI key
XTJLAIOQORFEOI-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
S Namciu et al.
Oncogene, 9(5), 1407-1416 (1994-05-01)
Overexpression of v-ski blocks the terminal differentiation of chicken erythroblasts, and in cooperation with v-sea causes transformation of these cells, indicating that c-ski may play a role in regulating either proliferation or differentiation in hematopoietic cells. We examined c-ski expression
B T French et al.
Cancer chemotherapy and pharmacology, 27(3), 171-177 (1990-01-01)
A variety of compounds inhibit the growth and induce differentiation of human promyelocytic leukemia (HL-60) cells. HL-60 subclones that lack the purine salvage enzyme hypoxanthine-guanine phosphoribosyltransferase (HGPRT) can also be induced to differentiate with purine analogs. Mechanisms by which purine
D S Gibboney et al.
Cancer chemotherapy and pharmacology, 25(3), 189-194 (1989-01-01)
A variety of purine analogs inhibit the growth and induce the differentiation of human promyelocytic leukemia (HL-60) cells that lack the purine salvage enzyme hypoxanthine-guanine phosphoribosyltransferase (HGPRT). Mechanisms by which purine analogs induce differentiation offer unique potential for cancer chemotherapy.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service