Skip to Content
Merck
All Photos(1)

Key Documents

E139

Sigma-Aldrich

BQ-3020

≥90%, powder

Synonym(s):

N-Acetyl-[Ala11,15]-Endothelin 1 fragment 6-21

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C96H140N20O25S
CAS Number:
Molecular Weight:
2006.32
MDL number:
UNSPSC Code:
12352200

Assay

≥90%

form

powder

color

white

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](C)NC(=O)[C@H](Cc2ccccc2)NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CC(O)=O)NC(=O)[C@H](CCSC)NC(=O)[C@H](CC(C)C)NC(C)=O)C(C)C)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N[C@@H](Cc4c[nH]c5ccccc45)C(O)=O

Gene Information

human ... EDN1(1906)

Amino Acid Sequence

Ac-Leu-Met-Asp-Lys-Glu-Ala-Val-Tyr-Phe-Ala-His-Leu-Asp-Ile-Ile-Trp

Biochem/physiol Actions

Selective ETB endothelin receptor agonist.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ihara et al.
Life sciences, 51(6), PL47-PL52 (1992-01-01)
A linear endothelin (ET) analog, N-acetyl-LeuMetAspLysGluAlaValTyrPheAlaHisLeu-AspIleIleTrp (BQ-3020), is highly selective for ETB receptors. BQ-3020 displaces [125I]ET-1 binding to ETB receptors (nonselective to ET isopeptides) in porcine cerebellar membranes (IC50: 0.2nM) at a concentration 4,700 times lower than that to ETA
A P Davenport et al.
Journal of cardiovascular pharmacology, 22 Suppl 8, S22-S25 (1993-01-01)
Our aim was to characterize and determine the function of endothelin (ET) receptor subtypes in human vascular tissue. Reverse transcriptase-polymerase chain reaction with nested oligonucleotide primers detected the presence of mRNA encoding both ETA and ETB receptors in the media
U Ekelund et al.
British journal of pharmacology, 112(3), 887-894 (1994-07-01)
1. This paper describes, in quantitative terms, the in vivo effects of two selective ETB-receptor agonists (IRL 1620 and BQ 3020) on vascular resistance (tone) in the following consecutive sections of the vascular bed of sympathectomized cat skeletal muscle: large-bore
Peter Johnström et al.
Experimental biology and medicine (Maywood, N.J.), 231(6), 736-740 (2006-06-03)
The endothelin (ET) receptor system has been shown to play a role in a number of vascular diseases. We have synthesized 18F-and 11C-labeled radioligands to enable in vivo imaging of the fundamental processes involved in ET receptor pharmacology in normal
Xiao Tao et al.
Science (New York, N.Y.), 326(5960), 1668-1674 (2009-12-19)
Inward-rectifier potassium (K+) channels conduct K+ ions most efficiently in one direction, into the cell. Kir2 channels control the resting membrane voltage in many electrically excitable cells, and heritable mutations cause periodic paralysis and cardiac arrhythmia. We present the crystal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service