Skip to Content
Merck
All Photos(1)

Key Documents

E0156

Sigma-Aldrich

Anti-Estrogen-Related Receptor β antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ERRβ, Anti-NR3B2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1 μg/mL using human liver, hepatocytes

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ESRRB(2103)
mouse ... Esrrb(26380)
rat ... Esrrb(299210)

Immunogen

synthetic peptide corresponding to the internal domain of human estrogen-related receptor β, conjugated to KLH. The immunizing peptide has 93% homology with the rat and mouse gene.

Application

Anti-Estrogen-Related Receptor β antibody produced in rabbit was used for immunocytochemistry of neurons from mouse model of glaucoma at a working dilution of 1:500. It was also used for immunohistochemistry of mouse brain sections.

Biochem/physiol Actions

Estrogen-Related Receptors (ERR) are hormone receptors that bind estrogen response elements that in turn bind specific DNA sequences and regulate transcription. Three ERRs are known, alpha, beta and gamma, with highly conserved ligand and DNA binding sequences. ERRβ is selectively expressed in brain and is an important modulator of energy balance, food intake, neuropeptide levels and response to stress.

Physical form

Solution in phosphate buffered saline, containing 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Crish et al.
Neuroscience, 229, 55-70 (2012-11-20)
Failure of anterograde transport to distal targets in the brain is a common feature of neurodegenerative diseases. We have demonstrated in rodent models of glaucoma, the most common optic neuropathy, early loss of anterograde transport along the retinal ganglion cell
William A Alaynick
Mitochondrion, 8(4), 329-337 (2008-04-01)
Lipid metabolism is a continuum from emulsification and uptake of lipids in the intestine to cellular uptake and transport to compartments such as mitochondria. Whether fats are shuttled into lipid droplets in adipose tissue or oxidized in mitochondria and peroxisomes
Francisco M Nadal-Nicolás et al.
Zoological research, 44(1), 226-248 (2023-01-04)
Univocal identification of retinal ganglion cells (RGCs) is an essential prerequisite for studying their degeneration and neuroprotection. Before the advent of phenotypic markers, RGCs were normally identified using retrograde tracing of retinorecipient areas. This is an invasive technique, and its
María Angeles Real et al.
Neuroscience letters, 438(1), 48-53 (2008-05-13)
Recently, a new nuclear receptor subfamily has been identified and referred to as estrogen-related receptors. This new group shares sequence similarity, target genes, co-regulatory proteins, and action sites with the estrogen receptors; however, natural estrogens are not estrogen-related receptors ligands.
Silmara de Lima et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(23), 9149-9154 (2012-05-23)
The mature optic nerve cannot regenerate when injured, leaving victims of traumatic nerve damage or diseases such as glaucoma with irreversible visual losses. Recent studies have identified ways to stimulate retinal ganglion cells to regenerate axons part-way through the optic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service