Key Documents
DWK261352254
DURAN® culture tubes
new
glass tube diam. × H 16 mm × 150 mm, GL 18 thread, tube capacity 20 mL, with cap, borosilicate glass 3.3 tube
About This Item
UNSPSC Code:
41121706
material
PBT cap
PTFE seal
borosilicate glass 3.3 tube
Joint
joint threaded neck
feature
autoclavable
with cap
packaging
pack of 50 ea
manufacturer/tradename
DWK Life Sciences DURAN® 261352254
glass tube diam. × H
16 mm × 150 mm
tube capacity
20 mL
fitting
GL 18 thread
suitability
suitable for (culture)
General description
The in vitro cultivation of animal and plant cells in a nutrient medium is an essential part of today′′s research in the field of microbiology. These cultivation approaches can take place at different scales - the vessels are selected accordingly. For test runs, rather small vessels such as culture tubes are used. The original DURAN® culture tubes are distinguished by their uniform wall thickness distribution, minimal thermal expansion and the resulting high resistance to temperature changes. They are therefore particularly suitable for work at very high temperatures and rapid temperature changes.
Application
In vitro cell culture
Features and Benefits
  • Manufactured from borosilicate glass 3.3
  • With DIN thread and screw cap from PBT for airtight sealing
  • High tightness due to PTFE-coated silicone seal
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)