Skip to Content
Merck
All Photos(4)

Key Documents

DNB200

Sigma-Aldrich

GenElute Stool DNA Isolation Kit

greener alternative
Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600
NACRES:
NA.52

greener alternative product characteristics

Designing Safer Chemicals
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

shipped in

ambient

storage temp.

room temp

General description

Sigma′s GenElute Stool DNA Isolation Kit provides a convenient and rapid method to isolate total DNA from fresh or frozen stool samples. The kit can also be used to isolate DNA from preserved stool samples. The universal protocol conveniently allows for the isolation of total genomic DNA from all the various microorganisms and host cells found in the stool sample simultaneously. The kit removes all traces of humic acid using the provided Bead Tubes and a combination of chemical and physical homogenization and lysis. A simple and rapid spin column procedure is then used to further purify the DNA. The purified DNA is of the highest quality and is fully compatible with downstream PCR applications, as all humic acid substances and PCR inhibitors are removed during the isolation.

  • Simultaneous isolation of both host DNA and microbial DNA (universal protocol)
  • Eliminates PCR inhibitors including humic acid
  • High quality DNA for sensitive downstream applications

Application

  • PCR
  • Southern Blot Analysis
  • Sequencing
  • Microarray Analysis

Features and Benefits

  • Fast and easy processing - Rapid isolation of high-quality, ready-to-use DNA in 30 minutes.
  • No organic extraction or alcohol precipitation - Phenol is not required for the isolation
  • Universal procedure - Simultaneous isolation of both host DNA and microbial DNA
  • High yields - Consistent, high yields of inhibitor-free DNA up to 50 kb plus
  • Remove all inhibitors - Eliminates all PCR inhibitors including humic acid

Unit Definition

Preps

Storage and Stability

All solutions should be kept tightly sealed and stored at room temperature. These reagents should remain stable for at least 1 year in their unopened containers.

Other Notes

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has Inherently Safer Chemistry, compared to the standard use of phenol and chloroform to perform DNA extractions.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Doina Georgescu et al.
International journal of general medicine, 15, 5513-5531 (2022-06-16)
Gallstone disease (GSD) is more commonly presented in aged people. The purpose of the study was to explore the insights of metabolic performance of bacterial species from gut microbiota as well as the clinical background in middle-aged and elderly patients
Qi-da He et al.
Evidence-based complementary and alternative medicine : eCAM, 2019, 6184205-6184205 (2020-01-18)
In Traditional Chinese Medicine (TCM), moxibustion had been used for thousands of years. Many clinical case reports and scientific studies had proved that moxibustion had a good effect in treating acute gastric ulcer (AGU). Some studies had shown that the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service