Skip to Content
Merck
All Photos(2)

Key Documents

D8519

Sigma-Aldrich

1,2-Distearoyl-rac-glycerol

≥99%

Synonym(s):

1,2-Dioctadecanoyl-rac-glycerol, 1,2-Distearin, DG(18:0/18:0/0:0)[rac], NSC 75181, Octadecanoic acid 1,1′-[1-(hydroxymethyl)-1,2-ethanediyl] ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

≥99%

form

powder

impurities

1,3-isomer, essentially free

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3

InChI key

UHUSDOQQWJGJQS-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service