Skip to Content
Merck
All Photos(1)

Key Documents

D8259

Sigma-Aldrich

Deoxyribonucleic acid from Micrococcus luteus (lysodeikticus)

Type XI, Highly polymerized

Synonym(s):

DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
eCl@ss:
32160414
NACRES:
NA.51

type

Type XI

form

solid

storage temp.

2-8°C

InChI

1S/C15H31N3O13P2/c16-13-1-7(20)11(28-13)5-25-32(21,22)31-9-3-15(18)29-12(9)6-26-33(23,24)30-8-2-14(17)27-10(8)4-19/h7-15,19-20H,1-6,16-18H2,(H,21,22)(H,23,24)

InChI key

AWBASQCACWFTGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

DNAs are useful for genomic analysis, including PCR, library construction in bacteriophage lambda, and in heterologous pre-hybridization and hybridization protocols. Micrococcus luteus DNA contains a high G-C content (72%) versus other DNAs like Clostridium perfringens DNA, G + C (27%), making it useful in studies that compare the effects of base composition on DNA properties.
The DNAs are particularly useful for genomic analysis, including PCR, library construction in bacteriophage lambda, and in heterologous pre-hybridization and hybridization protocols.

Quality

Contains 2-4% protein

Preparation Note

Prepared by procedure of Marmur, J., J. Mol. Biol., 3, 208 (1961).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maarten Holkers et al.
Nature methods, 11(10), 1051-1057 (2014-08-26)
Engineered sequence-specific nucleases and donor DNA templates can be customized to edit mammalian genomes via the homologous recombination (HR) pathway. Here we report that the nature of the donor DNA greatly affects the specificity and accuracy of the editing process
Manolis Kellis et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(17), 6131-6138 (2014-04-23)
With the completion of the human genome sequence, attention turned to identifying and annotating its functional DNA elements. As a complement to genetic and comparative genomics approaches, the Encyclopedia of DNA Elements Project was launched to contribute maps of RNA
Zachary D Smith et al.
Nature, 511(7511), 611-615 (2014-08-01)
In mammals, cytosine methylation is predominantly restricted to CpG dinucleotides and stably distributed across the genome, with local, cell-type-specific regulation directed by DNA binding factors. This comparatively static landscape is in marked contrast with the events of fertilization, during which
Hasan Yardimci et al.
Nature structural & molecular biology, 21(1), 20-25 (2014-01-07)
In G1, two copies of the MCM2-7 helicase are recruited to each origin of replication. Whereas recruitment of the first MCM2-7 is likely to be analogous to the loading of sliding clamps around DNA, how the second MCM2-7 complex is
Garrett Hellenthal et al.
Science (New York, N.Y.), 343(6172), 747-751 (2014-02-18)
Modern genetic data combined with appropriate statistical methods have the potential to contribute substantially to our understanding of human history. We have developed an approach that exploits the genomic structure of admixed populations to date and characterize historical mixture events

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service