Skip to Content
Merck
All Photos(1)

Key Documents

D7257

Sigma-Aldrich

6-(γ,γ-Dimethylallylamino)purine riboside

Synonym(s):

2iP Riboside, 6-(3,3-Dimethylallylamino)purine riboside, N6-(γ,γ-Dimethylallyl)adenosine, N6-(2-Isopentenyl)adenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21N5O4
CAS Number:
Molecular Weight:
335.36
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

storage temp.

−20°C

SMILES string

C\C(C)=C\CNc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Rajabi et al.
Nucleic acid therapeutics, 21(5), 355-358 (2011-10-19)
N(6)-Isopentenyladenosine (iPA), a member of the cytokinin family of plant hormones, exerts remarkable inhibition on tumor cell proliferation and apoptosis in several tumor cell lines. In this study, we report that iPA is able to inhibit the proliferation and promotes
Danuše Tarkowská et al.
Plant science : an international journal of experimental plant biology, 187, 105-112 (2012-03-13)
The putative role of cytokinins in processes leading to reproductive development of plants was investigated by analysing the shoot apical parts of a winter cultivar of oilseed rape (Brassica napus L. var. oleifera, cv. Górczański). The endogenous cytokinin levels were
Michael Behr et al.
Molecular plant-microbe interactions : MPMI, 25(8), 1073-1082 (2012-07-04)
When inoculated onto maize leaves at the onset of senescence, the hemibiotroph Colletotrichum graminicola causes green islands that are surrounded by senescing tissue. Taking advantage of green islands as indicators of sites of the establishment of successful infection and of
Zhong-Bao Yang et al.
Journal of experimental botany, 63(8), 3109-3125 (2012-03-01)
Aluminium (Al) toxicity and drought are two major factors limiting common bean (Phaseolus vulgaris) production in the tropics. Short-term effects of Al toxicity and drought stress on root growth in acid, Al-toxic soil were studied, with special emphasis on Al-drought
Veronika Reiter et al.
Nucleic acids research, 40(13), 6235-6240 (2012-03-17)
The unusual cyclin-dependent protein kinase 5 (CDK5) was discovered based on its sequence homology to cell cycle regulating CDKs. CDK5 was found to be active in brain tissues, where it is not involved in cell cycle regulation but in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service