Skip to Content
Merck
All Photos(1)

Key Documents

D7159

Sigma-Aldrich

2′,3′-Dideoxycytidine 5′-triphosphate sodium salt

≥90% (HPLC)

Synonym(s):

ddCTP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16N3O12P3
CAS Number:
Molecular Weight:
451.16
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

biological source

synthetic (organic)

Assay

≥90% (HPLC)

form

powder

solubility

water: 1 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].NC1=NC(=O)N(C=C1)[C@H]2CC[C@@H](COP([O-])(=O)OP(O)(=O)OP(O)(O)=O)O2

InChI

1S/C9H16N3O12P3.Na/c10-7-3-4-12(9(13)11-7)8-2-1-6(22-8)5-21-26(17,18)24-27(19,20)23-25(14,15)16;/h3-4,6,8H,1-2,5H2,(H,17,18)(H,19,20)(H2,10,11,13)(H2,14,15,16);/q;+1/p-1/t6-,8+;/m0./s1

InChI key

LOOMQRBEBYCFPP-QDOHZIMISA-M

Application

Complexes of ddCTP with DNA polymerase β were studied by x-ray crystallography, with implications for inhibitory activity against HIV reverse transcriptase.

Biochem/physiol Actions

Inhibitor of DNA polymerase I-catalyzed chain elongation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Georges Massiot et al.
Journal of natural products, 68(7), 979-984 (2005-07-26)
Five new acylphloroglucinol derivatives, mahureones A-E (1, 3-6), have been isolated from the leaves of Mahurea palustris, and their structures determined by spectroscopic means. During the isolation process, several byproducts (7-9) were formed by reaction of one of the isoprenyl
Yaligara Veeranagouda et al.
BMC genomics, 15, 429-429 (2014-06-06)
Bacteroides fragilis is a Gram-negative anaerobe that is normally a human gut commensal; it comprises a small percentage of the gut Bacteroides but is the most frequently isolated Bacteroides from human infections. Identification of the essential genes necessary for the
Michal R Szymanski et al.
The EMBO journal, 34(14), 1959-1970 (2015-06-10)
The human DNA polymerase gamma (Pol γ) is responsible for DNA replication in mitochondria. Pol γ is particularly susceptible to inhibition by dideoxynucleoside-based inhibitors designed to fight viral infection. Here, we report crystal structures of the replicating Pol γ-DNA complex
Nutjaree Jeenduang et al.
Clinica chimica acta; international journal of clinical chemistry, 442, 36-43 (2015-01-17)
Cardiovascular disease (CVD) is one of the major causes of death worldwide. Numerous genetic risk factors in lipid metabolism, including mutations of LDLR, APOB, and PCSK9, as well as polymorphisms of CETP and APOE, have been found to associate with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service