Skip to Content
Merck
All Photos(1)

Key Documents

D5766

Sigma-Aldrich

5,5-Dimethyl-1-pyrroline N-oxide

≥97%

Synonym(s):

DMPO

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO
CAS Number:
Molecular Weight:
113.16
Beilstein:
107603
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥97% (GC)
≥97%

form

solid

refractive index

n20/D 1.496 (lit.)

bp

75 °C/0.4 mmHg (lit.)

mp

25-29 °C (lit.)

density

1.015 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

−20°C

SMILES string

CC1(C)CCC=[N+]1[O-]

InChI

1S/C6H11NO/c1-6(2)4-3-5-7(6)8/h5H,3-4H2,1-2H3

InChI key

VCUVETGKTILCLC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

5,5-Dimethyl-1-pyrroline N-oxide has been used as a spin trapping reagent to detect free radicals in electron paramagnetic resonance (EPR) based reactions.[1][2]
Neuroprotective agent; nitric oxide spin trap. Used to study radicals formed by enzymatic acetaldehyde oxidation.
Neuroprotective agent; nitric oxide spin trap. Used to study radicals formed by enzymatic acetaldehyde oxidation. Incubation of lymphocytes with DMPO decreased DNA damage by NiCl2.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wan-Ook Ji et al.
Scientific reports, 9(1), 19837-19837 (2019-12-29)
Medical treatment utilizing non-thermal plasma is based on the production of reactive oxygen species (ROS) and their interactions with biomatters. On the basis of empirical data from practices, plasma treatment has been planned with regard to the setup of a
Xiaoguang Duan et al.
Small (Weinheim an der Bergstrasse, Germany), 11(25), 3036-3044 (2015-03-20)
Sulfur and nitrogen co-doped reduced graphene oxide (rGO) is synthesized by a facile method and demonstrated remarkably enhanced activities in metal-free activation of peroxymonosulfate (PMS) for catalytic oxidation of phenol. Based on first-order kinetic model, S-N co-doped rGO (SNG) presents
Mitochondrial O 2 and H 2O 2 mediate glucose deprivation-induced cytotoxicity and oxidative stress in human cancer cells
Ahmad IM, et al.
The Journal of biological chemistry (2004)
Nanoparticle-induced oxidation of corona proteins initiates an oxidative stress response in cells
Jayaram DT, et al.
Nanoscale, 9(22), 7595-7601 (2017)
Patrick T Kang et al.
Free radical biology & medicine, 53(4), 962-973 (2012-05-29)
Complex I is a critical site of O(2)(•-) production and the major host of reactive protein thiols in mitochondria. In response to oxidative stress, complex I protein thiols at the 51- and 75-kDa subunits are reversibly S-glutathionylated. The mechanism of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service