Skip to Content
Merck
All Photos(1)

Key Documents

D4376

Sigma-Aldrich

Deuterium chloride solution

35% in D2O

Synonym(s):

Hydrochloric acid-d solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
DCl
CAS Number:
Molecular Weight:
37.47
EC Number:
MDL number:
UNSPSC Code:
12352200

Pricing and availability is not currently available.

isotopic purity

99 atom % D

concentration

35% in D2O

mass shift

M+1

SMILES string

Cl[2H]

InChI

1S/ClH/h1H/i/hD

InChI key

VEXZGXHMUGYJMC-DYCDLGHISA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service