Skip to Content
Merck

Skip To

D2192

Desthiobiotin polyethyleneoxide Iodoacetamide

≥90% (HPLC)

Synonym(s):

Desthiobiotin PEO iodoacetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H38IN5O6
Molecular Weight:
583.46
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90% (HPLC)

form

powder

solubility

H2O: 10 mg/mL

storage temp.

2-8°C

SMILES string

CC1NC(=O)NC1CCCCCC(=O)NCCC(=O)NCCOCCOCCNC(=O)CI

InChI

1S/C21H38IN5O6/c1-16-17(27-21(31)26-16)5-3-2-4-6-18(28)23-8-7-19(29)24-9-11-32-13-14-33-12-10-25-20(30)15-22/h16-17H,2-15H2,1H3,(H,23,28)(H,24,29)(H,25,30)(H2,26,27,31)

InChI key

DBOWCCSTSCXTEL-UHFFFAOYSA-N

Application

Especially useful for cysteine labeling in many proteomics applications such as peptide mapping and mass spectrometry.

Features and Benefits

  • Easily eluted from streptavidin or avidin affinity matrices under mild conditions
  • Incorportates a 16 atom hydrophilic spacer
  • Typically coupled to sulfhydryl groups at pH 7.5 - 8.5

Other Notes

Sulfhydryl specific, water soluble biotinylation reagent.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Michael B Goshe et al.
Journal of proteome research, 2(2), 153-161 (2003-04-29)
The ability to identify and quantitate integral membrane proteins is an analytical challenge for mass spectrometry-based proteomics. The use of surfactants to solubilize and facilitate derivatization of these proteins can suppress peptide ionization and interfere with chromatographic separations during microcapillary
Advances in proteome analysis by mass spectrometry.
T J Griffin et al.
The Journal of biological chemistry, 276(49), 45497-45500 (2001-10-05)
T P Conrads et al.
Analytical chemistry, 73(9), 2132-2139 (2001-05-17)
We describe the combined use of 15N-metabolic labeling and a cysteine-reactive biotin affinity tag to isolate and quantitate cysteine-containing polypeptides (Cys-polypeptides) from Deinococcus radiodurans as well as from mouse B16 melanoma cells. D. radiodurans were cultured in both natural isotopic



Questions

Reviews

No rating value

Active Filters