Skip to Content
Merck
All Photos(1)

Key Documents

D1554

Sigma-Aldrich

3,3′-Dipentyloxacarbocyanine iodide

≥95%

Synonym(s):

DiOC5(3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H33IN2O2
CAS Number:
Molecular Weight:
544.47
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Pricing and availability is not currently available. Contact Technical Service

Assay

≥95%

SMILES string

[I-].CCCCCN1C(\Oc2ccccc12)=C\C=C\c3oc4ccccc4[n+]3CCCCC

Looking for similar products? Visit Product Comparison Guide

Application

Probe for measuring membrane potential and localization of endoplasmic reticulum

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K G Lazzari et al.
The Journal of biological chemistry, 265(19), 10959-10967 (1990-07-05)
The chemotactic peptide formylmethionyl-leucyl-phenylalanine (fMLP), at concentrations below 10(-9) M, elicits a sustained increase in the human neutrophil's membrane potential within 10 s of its addition. This hyperpolarization, detected with the fluorescent cationic potentiometric probes, 3,3'-dipentyloxacarbocyanine (diO-C5-(3)), and 1,1'-dipropyl-3,3,3',3'-tetramethylindocarbocyanine iodide
M Terasaki et al.
Cell, 38(1), 101-108 (1984-08-01)
Certain fluorescent dyes, previously reported to localize mitochondria, when used at higher concentrations also localize a continuous net-like structure in both living and glutaraldehyde-fixed cells. A similar reticular structure can be detected by phase-contrast microscopy and whole-mount electron microscopy in
C Haanen et al.
Histochemistry, 84(4-6), 609-613 (1986-01-01)
The measurement of time correlated intracellular mitochondrial staining with 3,3'-dipentyloxacarbocyanine [Di-O-C5(3)] appeared of interest to define the optimal staining conditions. Mitochondrial staining of lymphocytes, monocytes and granulocytes results in different fluorescence signals, related to the numbers of mitochondria, that are
A Favel et al.
The Journal of antimicrobial chemotherapy, 43(2), 227-232 (2001-03-17)
A flow cytofluorometric susceptibility test (FCST) was used for rapid determination of the susceptibility of Candida lusitaniae isolates to amphotericin B. The test is based on the decrease in fluorescence intensity of cells stained with 3,3'-dipentyloxacarbocyanine iodide (DiOC5(3)), a membrane
Konrad Kleszczyński et al.
Toxicology and applied pharmacology, 234(3), 300-305 (2008-11-26)
Perfluorinated (aliphatic) acids (PFAs) and congeners have many applications in various industrial fields and household for decades. Years later they have been detected in wildlife and this has spurred interest in environmental occurrence as well as influencing living organisms. PFAs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service