Skip to Content
Merck
All Photos(1)

Key Documents

D144

Sigma-Aldrich

DALDA

>96%, solid

Synonym(s):

[D-Arg2, Lys4]-Dermorphin-(1-4)-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H44N8O6
CAS Number:
Molecular Weight:
612.72
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>96%

form

solid

color

white

storage temp.

−20°C

SMILES string

NCCCCC(NC(=O)C(Cc1ccccc1)NC(=O)C(CCCNC(N)=N)NC(=O)C(N)Cc2ccc(O)cc2)C(N)=O

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Tyr-D-Arg-Phe-Lys-NH2

Biochem/physiol Actions

Highly selective μ opioid receptor agonist.

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Min Zhao et al.
The Journal of pharmacology and experimental therapeutics, 302(1), 188-196 (2002-06-18)
Recent studies suggest that delta-opioid receptors play a role in the development of opioid tolerance and led us to hypothesize that highly selective mu-opioid agonists may produce less tolerance. H-2',6'-dimethyltyrosine-D-Arg-Phe-Lys-NH(2) ([Dmt(1)]DALDA) has extraordinary selectivity for mu-receptors (K(i)(delta)/K(i)(mu) > 14,000). Daily
Damien J Keating et al.
The Journal of physiology, 555(Pt 2), 489-502 (2004-01-16)
Before the preganglionic regulation of the adrenal medulla is established, hypoxia acts directly on the chromaffin cells to evoke the secretion of catecholamines. This direct action of hypoxia is suppressed by the gradual development of the preganglionic innervation and we
Pal Riba et al.
Current medicinal chemistry, 9(1), 31-39 (2002-02-28)
The clinical effectiveness of morphine is limited by several side effects, including the development of tolerance and dependence. Most of these side effects are believed to be mediated by central opioid receptors; therefore, hydrophilic opioids, which don't cross the blood-brain
Yong Ben et al.
British journal of pharmacology, 143(8), 987-993 (2004-11-24)
Previously, we reported that H-2',6'-dimethyltyrosine [Dmt(1)]-d-Arg-Phe-Lys-NH(2) (DALDA), an analogue of the naturally occurring opioid peptide dermorphin, is a highly potent and selective mu receptor agonist with low cross-tolerance to morphine. In the present study, we investigated the effect of treating
Haibao Wan et al.
Rapid communications in mass spectrometry : RCM, 17(6), 538-546 (2003-03-07)
The synthetic peptide [Dmt(1)]DALDA (Dmt-D-Arg-Phe-Lys-NH(2); Dmt = 2',6'-dimethyltyrosine; 'super-DALDA') is a mu opioid-receptor agonist. On-line liquid chromatography/quadrupole time-of-flight mass spectrometry and the corresponding stable isotope-incorporated synthetic peptide internal standard were used to quantify [Dmt(1)]DALDA that had been extracted from ovine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service