Skip to Content
Merck
All Photos(3)

Documents

D1429

Sigma-Aldrich

2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate

99% purity (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H19N2O5Cl
CAS Number:
Molecular Weight:
378.81
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

2-[4-(Dimethylamino)styryl]-N-methylbenzoxazolium perchlorate, 99% (TLC)

Assay

99% (TLC)

form

powder

color

dark red

solubility

DMSO: 10 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[O-]Cl(=O)(=O)=O.CN(C)c1ccc(cc1)\C=C\c2o[n+](C)c3ccccc23

InChI

1S/C18H19N2O.ClHO4/c1-19(2)15-11-8-14(9-12-15)10-13-18-16-6-4-5-7-17(16)20(3)21-18;2-1(3,4)5/h4-13H,1-3H3;(H,2,3,4,5)/q+1;/p-1

InChI key

FDPJLIFXRNZDJR-UHFFFAOYSA-M

Application

DNA, RNA stain with advantageous spectral properties for cytometric applications.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Appl. Fluoresc. Technol., 5, 3-3 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service