Skip to Content
Merck
All Photos(1)

Key Documents

D142

Sigma-Aldrich

4-Diphenylacetoxy-N-(2-chloroethyl)piperidine hydrochloride

solid

Synonym(s):

4-DAMP mustard hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24ClNO2 · HCl
CAS Number:
Molecular Weight:
394.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

color

white

solubility

DMSO: soluble (Solutions must be freshly prepared.)
aqueous base: unstable

SMILES string

Cl[H].ClCCN1CCC(CC1)OC(=O)C(c2ccccc2)c3ccccc3

Biochem/physiol Actions

Irreversible muscarinic acetylcholine receptor antagonist with essentially equivalent affinity for M1, M3, M4, and M5 receptors and much lower affinity for M2 receptors.

Caution

Hygroscopic

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F J Ehlert et al.
Life sciences, 62(17-18), 1659-1664 (1998-05-19)
Irreversible ligands are useful tools for investigating the function of receptor subtypes in various physiological processes. The mechanism for alkylation involves the formation of a reversible receptor complex followed by a covalent reaction. The extent of receptor alkylation is determined
E A Thomas et al.
Molecular pharmacology, 44(1), 102-110 (1993-07-01)
A functional role for the M2 muscarinic receptor in smooth muscle contraction was investigated in isolated guinea pig ileum. Contractile responses to the muscarinic agonist oxotremorine-M (oxo-M) were measured in isolated ilea that had been pretreated with histamine (0.32 microM)
N Watson et al.
European journal of pharmacology, 278(3), 195-201 (1995-05-24)
Muscarinic M2 receptors account for more than half the muscarinic receptor population in smooth muscles of a number of species and yet it is the smaller M3 receptor population that mediates contraction of many of these tissues. The role of
R S Ostrom et al.
The Journal of pharmacology and experimental therapeutics, 286(1), 234-242 (1998-07-10)
The ability of the M2 muscarinic receptor to inhibit the relaxant effects of forskolin and isoproterenol was investigated in bovine trachea. In most experiments, we measured contractile responses to oxotremorine-M in smooth muscle isolated from bovine trachea in which a
M Waelbroeck et al.
Biochemical pharmacology, 44(2), 285-290 (1992-07-22)
We demonstrated in this study that 4-DAMP [4-diphenylacetoxy-1-(2- chloroethyl) piperidine] mustard, which cyclizes to the aziridinium ion, behaved as a non-selective, non-competitive inhibitor of muscarinic receptors in rat brain cortex. It inactivated to the same extent the M1, M2 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service