D0634
1-Decanol
≥98%
Synonym(s):
n-Decyl alcohol, Alcohol C10
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)9OH
CAS Number:
Molecular Weight:
158.28
Beilstein:
1735221
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
vapor density
4.5 (vs air)
vapor pressure
1 mmHg ( 70 °C)
8.25 mmHg ( 100 °C)
Assay
≥98%
autoignition temp.
550 °F
refractive index
n20/D 1.437 (lit.)
bp
231 °C (lit.)
mp
5-7 °C (lit.)
density
0.829 g/mL at 25 °C (lit.)
SMILES string
CCCCCCCCCCO
InChI
1S/C10H22O/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3
InChI key
MWKFXSUHUHTGQN-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
Product No.
Description
Pricing
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service