Skip to Content
Merck
All Photos(5)

Key Documents

CWA-1158

Sigma-Aldrich

Anti-Human CD276/B7-H3 (376.96) ColorWheel® Dye-Ready mAb

greener alternative

Synonym(s):

CD276 antigen;4Ig-B7-H3;B7 homolog 3;B7-H3;Costimulatory molecule

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product CWA-1158 is not currently sold in your country. Contact Technical Service

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

376.96, monoclonal

product line

ColorWheel®

form

lyophilized

mol wt

calculated mol wt 57.24 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

Protein ID accession no.

UniProt accession no.

compatibility

for use with ColorWheel® Dyes (Required, (Sold Separately))

greener alternative category

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD276(80381)

General description

ColorWheel® technology is a novel and proprietary method of creating your own antibody and dye combinations for use in flow cytometry. By incubating any ColorWheel® antibody with any ColorWheel® dye, researchers can quickly and simply produce primary conjugated antibodies for use in single color or multicolor flow cytometry analysis. Each ColorWheel® antibody and dye is lyophilized for long-term storage, allowing for simplicity and flexibility without compromise. ColorWheel® technology requires both ColorWheel® antibody and ColorWheel® dye for Flow Cytometry application.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This product is Preservative-free, lyophilized product for enhanced stability and allow for ambient shipping and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Specificity

Clone 376.96 is a mouse monoclonal antibody that detects CD276/B7-H3. It targets an epitope within the extracellular domain.

Immunogen

Cultured COLO 38 human melanoma cells.

Application

Quality Control Testing

Evaluated by Flow Cytometry in SK-OV-3 cells.

Flow Cytometry Analysis: Staining of one million SK-OV-3 cells was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1158, Anti-Human CD276/B7-H3 (376.96) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye or an equivalent amount of Cat. No. CWA-1016 Mouse IgG2a isotype control and Cat. No. CWDS-PE ColorWheel® Antibody-Ready Phycoerythrin (PE) Dye.

Tested Applications

Flow Cytometry Analysis: Staining of one million SK-OV-3 cells was performed using 5 µL of a 1:1 mixture of Cat. No. CWA-1158, Anti-Human CD276/B7-H3 (376.96) ColorWheel® Dye-Ready mAb and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye or a 1:1 mixture of an equivalent amount of Cat. No. CWA-1016 Mouse IgG2a isotype control and Cat. No. CWDS-APC ColorWheel® Antibody-Ready Allophycocyanin (APC) Dye.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Compatibility

Requires ColorWheel® Dye (Sold Separately)

Target description

CD276 antigen (UniProt: Q5ZPR3; also known as 4Ig-B7-H3, B7 homolog 3, B7-H3, Costimulatory molecule, CD276) is encoded by the CD276 (also known as B7H3, PSEC0249, UNQ309/PRO352) gene (Gene ID: 80381) in human. CD276 is a single-pass type I membrane protein that is member of B7 immunoregulatory family. It is synthesized with a signal peptide (aa 1-28), which is subsequently cleaved off to produce the mature form, which contains an extracellular domain (aa 29-466), a short transmembrane domain (aa 467-487), and a cytoplasmic domain (aa 488-534). It participates in the regulation of T-cell-mediated immune response. It is a ubiquitous protein but is undetectable in peripheral blood lymphocytes or granulocytes and is only weakly expressed in resting monocytes. It is also expressed in dendritic cells derived from monocytes and in epithelial cells of sinonasal tissue, extravillous trophoblast cells and Hofbauer cells of the first trimester placenta and term placenta. It plays a key role in providing the placenta and fetus with a suitable immunological environment throughout pregnancy. Higher expression of CD276 has been shown in a variety of cancer types, including breast, pancreatic, and ovarian cancer and its expression has been correlated with poor survival in ovarian cancer subjects. It is suggested to play a protective role in tumor cells by inhibiting natural-killer mediated cell lysis. Its levels are also reported to be up-regulated in cells that mediate rejection of human transplants. This ColorWheel® Dye-Ready monoclonal antibody was developed using our Cat. No. MABC1731, Anti-B7-H3 (CD276) Antibody, clone 376.96. (Ref.: Fauci, JM., et al. (2014). Gynecologic Oncology 132(1); 203-210; Chen, Y-W., et al. (2008). Curr. Can. Drug Targets. 8(5); 404-413).

Physical form

Lyophilized from PBS containing D-Mannitol and Sucrose, normal appearance is a dried pellet. Reconstituted antibody solution is stable and functional as assessed by functional testing. Contains no biocide or preservatives, such as azide.

Reconstitution

1.0 mg/mL after reconstitution at 25 µL in PBS. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C. Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial. Reconstitute each vial by adding 25 µL PBS. Protect from light.

Legal Information

ColorWheel is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service