Skip to Content
Merck
All Photos(1)

Documents

CPS1A300

Sigma-Aldrich

Chemiluminescent Peroxidase Substrate-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.83

form

liquid

usage

sufficient for 300 mL substrate

storage temp.

2-8°C

Application

Chemiluminescent Peroxidase Substrate can be used for the highly sensitive detection of peroxidase labeled material in a variety of Western blotting applications. This substrate is an enhanced luminol product with a stabilized peroxide buffer solution that provides femtogram sensitivity with minimal background interference.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yeqing Fang et al.
International journal of molecular medicine, 48(2) (2021-07-03)
Myocardial ischemia triggers an inflammatory reaction and oxidative stress that increases apoptosis of myocardiocytes. It has been evidenced that tanshinone‑IIA (Tan‑IIA) protects against heart failure post‑myocardial infarction via inhibition of the apoptotic pathway. The purpose of the present study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service