CLS432087
Extension Collar, for Corning® CoolBox™ 2XT System
purple
Synonym(s):
Corning Coolcell accessory
Select a Size
€136.00
Select a Size
About This Item
€136.00
Recommended Products
packaging
pkg of 1
manufacturer/tradename
Corning 432087
L × W × H
21.0 cm × 14.3 cm × 7.0 cm, Internal Dimensions
26.5 cm × 20.0 cm × 7.0 cm, External Dimensions
color
purple
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service