Key Documents
CLS432075
Corning® ThermalTray
HP high profile for water bath
Synonym(s):
lab equipment accessory
Select a Size
1 EA
€427.00
€427.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
€427.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.12
€427.00
Please contact Customer Service for Availability
feature
HP high profile for water bath
packaging
pkg of 1
manufacturer/tradename
Corning 432075
L × W × H
28.0 cm × 14.0 cm × 8.6 cm
General description
Corning ThermalTray thermo-conductive platforms support CoolRack and CoolSink sample modules in liquid temperature sources such as melting ice, water baths and liquid nitrogen. Made of the same highly conductive alloy as CoolRack and CoolSink modules, ThermalTray platforms conduct the source temperature to the CoolRack or CoolSink and, ultimately, to your samples.
  • Stable, sturdy design makes them ideal for processing temperature-sensitive samples in melted ice baths or liquid nitrogen.
  • All modules may be autoclaved, high heat sterilized or decontaminated with bleach, alcohol or other disinfectants or lab detergents.
Legal Information
Corning is a registered trademark of Corning, Inc.
ThermalTray is a trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)